Skip to Content
Merck
All Photos(4)

Documents

C4695

Sigma-Aldrich

CHAPSO

BioXtra

Synonym(s):

3-([3-Cholamidopropyl]dimethylammonio)-2-hydroxy-1-propanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H58N2O8S
CAS Number:
Molecular Weight:
630.88
Beilstein:
5842642
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

zwitterionic

Quality Level

product line

BioXtra

Assay

≥98.0% (TLC)

form

powder

mol wt

micellar avg mol wt 7000

aggregation number

11

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.1%

CMC

8 mM (20-25°C)
8 mM (micellar weight =9960)

mp

184-186 °C (lit.)

transition temp

cloud point 90 °C

solubility

H2O: 0.1 M, clear, colorless

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.01%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

[H][C@@]12C[C@H](O)CC[C@]1(C)[C@@]3([H])C[C@H](O)[C@]4(C)[C@]([H])(CC[C@@]4([H])[C@]3([H])[C@H](O)C2)[C@H](C)CCC(=O)NCCC[N+](C)(C)CC(O)CS([O-])(=O)=O

InChI

1S/C32H58N2O8S/c1-20(7-10-29(39)33-13-6-14-34(4,5)18-23(36)19-43(40,41)42)24-8-9-25-30-26(17-28(38)32(24,25)3)31(2)12-11-22(35)15-21(31)16-27(30)37/h20-28,30,35-38H,6-19H2,1-5H3,(H-,33,39,40,41,42)/t20-,21+,22-,23?,24-,25+,26+,27-,28+,30+,31+,32-/m1/s1

InChI key

GUQQBLRVXOUDTN-XOHPMCGNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chapso is a zwitterionic detergent derived from Chaps by the addition of a functional hydroxyl group. It is a nondenaturing zwitterionic detergent with characteristics similar to CHAPS, although it is more soluble due to a more polar head group.

Application

Chapso has been used in a study to assess methods for high-throughput crystallization of membrane proteins. It has also been used in a study to investigate the effects of detergents on the structure of rhomboid proteases in a lipid environment.
A nondenaturing zwitterionic detergent with characteristics similar to CHAPS, although it is more soluble due to a more polar head group. Useful for the solubilization of integral membrane proteins.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Incomplete Dialysis of Protein Samples Containing 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate May Lead to Erroneous Estimation of Histidine Content on Amino Acid Analysis
Nomura, K., et al.
Analytical Biochemistry, 290, 4-4 (2001)
Structure of Rhomboid Protease in a Lipid Environment
Vinothkumar, K., et al.
Journal of Molecular Biology, 407, 16-16 (2011)
Satoko Osawa et al.
The Journal of biological chemistry, 283(28), 19283-19292 (2008-05-16)
gamma-Secretase is an aspartic protease that hydrolyzes type I membrane proteins within the hydrophobic environment of the lipid bilayer. Using the CHAPSO-solubilized gamma-secretase assay system, we previously found that gamma-secretase activity was sensitive to the concentrations of detergent and phosphatidylcholine.
Rachna Ujwal et al.
Methods (San Diego, Calif.), 55(4), 337-341 (2011-10-11)
Crystallization of membrane proteins remains a significant challenge. For proteins resistant to the traditional approach of directly crystallizing from detergents, lipidic phase crystallization can be a powerful tool. Bicelles are an excellent medium for crystallizing membrane proteins in a lipidic
Jonathan D J Wrigley et al.
Journal of neurochemistry, 90(6), 1312-1320 (2004-09-03)
Gamma-secretase performs the final processing step in the generation of amyloid-beta (Abeta) peptides, which are believed to be causative for Alzheimer's disease. Presenilins (PS) are required for gamma-secretase activity and the presence of two essential intramembranous aspartates (D257 and D385)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service