Skip to Content
Merck

Skip To

220201

CHAPS

Molecular Biology Grade

Synonym(s):

CHAPS, 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C32H58N2O7S
CAS Number:
Molecular Weight:
614.88
UNSPSC Code:
12161703
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

CHAPS, Molecular Biology Grade,

grade

Molecular Biology

Quality Segment

description

Merck USA index - 14, 2044, zwitterionic

assay

≥98% (NMR)

form

crystalline powder

mol wt

micellar avg mol wt 6150

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated (hygroscopic)

aggregation number

10

color

white

CMC

6 - 10 mM, 6 mM (20-25°C)

mp

157 °C (315 °F)

transition temp

cloud point >100 °C

solubility

water: 1.0 M

foreign activity

DNase, RNase, and Protease, none detected

shipped in

ambient

storage temp.

15-25°C

SMILES string

C[C@H](CCC(=O)NCCC[N+](C)(C)CCCS([O-])(=O)=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)CC4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

InChI

1S/C32H58N2O7S/c1-21(8-11-29(38)33-14-6-15-34(4,5)16-7-17-42(39,40)41)24-9-10-25-30-26(20-28(37)32(24,25)3)31(2)13-12-23(35)18-22(31)19-27(30)36/h21-28,30,35-37H,6-20H2,1-5H3,(H-,33,38,39,40,41)/t21-,22?,23-,24-,25+,26+,27-,28+,30+,31+,32-/m1/s1

InChI key

UMCMPZBLKLEWAF-RFCNGIAKSA-N

General description

Conductivity (1.0 M, 24°C): <100 µmhos.
Non-denaturing zwitterionic detergent that combines features of bile salts and N-alkyl sulfobetaines. Better at solubilizing proteins than the structurally related carboxylic acid anions, and is more effective at breaking protein-protein interactions than either sodium cholate or TRITON X-100. Capable of solubilizing the opiate receptor to a state exhibiting reversible binding to opiates. Also capable of disaggregating P 450 to its monomeric form without denaturation. Has a small micellar molecular weight (6150 Daltons) and a high critical micellar concentration (6-10 mM). Can be removed from either gels or protein solutions by dialysis. Aggregation number: 4-14.
Zwitterionic detergent that combines features of bile salts and N-alkyl sulfobetaines. Better at solubilizing proteins than structurally-related carboxylic acid anions and is much more effective at breaking protein-protein interactions than either sodium cholate or TRITON X-100 Detergent. Capable of solubilizing the opiate receptor to a state exhibiting reversible binding of opiates. Also, capable of disaggregating cytochrome P450 to its monomeric form without denaturation. Has a small micellar molecular weight and a high critical micellar concentration and can be removed from either gels or protein solutions by dialysis across a membrane. Aggregation number: 4-14.

Application

CHAPS is a nondenaturing zwitterionic detergent for membrane biochemistry.
Useful for solubilizing membrane proteins and breaking protein-protein interactions. CHAPS′ small micellar molecular weight (6,150) and high critical micelle concentration (6-10 mM) allow it to be removed from samples by dialysis. It is also suitable for protein solubilization for isoelectric focusing and two-dimensional electrophoresis. CHAPS is commonly used for non-denaturing (without urea) IEF and has been shown to give excellent resolution of some subcellular preparations and plant proteins. Concentrations between 2-4% (w/v) are typically used in an IEF gel.

Preparation Note

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 6 months at 4°C.

Other Notes

Chen, Y., et al. 1992. Biochemistry 31, 2415.
Ofri, D., et al. 1992. J. Neurochem. 58, 628.
Ransom, R.W., et al. 1992. Biochem. Pharmacol. 43, 1823.
Simonds, W.F., et al. 1992. Proc. Natl. Acad. Sci. USA77, 4623.
Yannariello-Brown, J., and Weigel, P.H. 1992. Biochemistry31, 576.
Hjelmeland, L.M. 1980. Proc. Natl. Acad. Sci. USA77, 6368.
Simonds, W.F., et al. 1980. Proc. Natl. Acad. Sci. USA77, 4623.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Disclaimer

Toxicity: Harmful (C)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
198991.11662850500P
description

-

description

-

description

for biochemistry

description

Avanti Research - A Croda Brand

grade

Molecular Biology

grade

-

grade

-

grade

-

assay

≥98% (NMR)

assay

-

assay

-

assay

>99% (TLC)

form

crystalline powder

form

liquid

form

solid

form

powder

aggregation number

10

aggregation number

-

aggregation number

10

aggregation number

-

storage temp.

15-25°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

mp

157 °C (315 °F)

mp

-

mp

157 °C

mp

-


Still not finding the right product?

Explore all of our products under CHAPS, Molecular Biology Grade


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–3 of 3 Questions  
  1. CHAPS - is derived from what animal? Working on a USDA Permit Template. Bovine? or Mouse?

    1 answer
    1. This product has historically been derived from Bovine and/or Ovine origin. However, this information is lot specific and may change over time. To assure the most accurate information, please refer to the lot-specific Certificate of Origin.

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters