Skip to Content
MilliporeSigma

Skip To

A4036

Adenosine

BioReagent, suitable for cell culture

Synonym(s):

9-β-D-Ribofuranosyladenine, Adenine riboside, Adenine-9-β-D-ribofuranoside

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 g

Available to ship TODAYfromMILWAUKEE

$49.60
25 g

Available to ship TODAYfromMILWAUKEE

$159.00

About This Item

Empirical Formula (Hill Notation):
C10H13N5O4
CAS Number:
Molecular Weight:
267.24
NACRES:
NA.75
PubChem Substance ID:
eCl@ss:
32160413
UNSPSC Code:
12352207
EC Number:
200-389-9
MDL number:
Beilstein/REAXYS Number:
93029
Assay:
≥99% (HPLC)
Form:
powder

$49.60


Available to ship TODAYDetails


Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic (organic)

Quality Segment

product line

BioReagent

assay

≥99% (HPLC)

form

powder

technique(s)

cell culture | mammalian: suitable

mp

234-236 °C (lit.)

solubility

1 M NH4OH: 50 mg/mL, clear, colorless

storage temp.

2-8°C

SMILES string

Nc1ncnc2n(cnc12)[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O

InChI

1S/C10H13N5O4/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(18)6(17)4(1-16)19-10/h2-4,6-7,10,16-18H,1H2,(H2,11,12,13)/t4-,6-,7-,10-/m1/s1

InChI key

OIRDTQYFTABQOQ-KQYNXXCUSA-N

General description

The nucleoside adenosine consists of the purine adenine and the sugar ribose. Thus, it is also referred as adenine-D-ribose. Adenosine is an important molecule that forms nucleotides, which is a part of nucleic acids forming the DNA and RNA. The nucleotides include: adenosine monophosphate, adenosine diphosphate and adenosine triphosphate.

Application

Adenosine has been used as a supplement in DMEM (Dulbecco′s modified Eagle medium):
  • to evaluate the outgrowth of the interspecies somatic cell nucleus transfer (iSCNT)-derived blastocysts
  • to maintain the CGR8 ES (germ-line competent cell line) cells
  • for plasmid transfection and cloning of embryonic stem cells

Biochem/physiol Actions

Endogenous neurotransmitter at adenosine receptors. Cardioprotective effects may relate to activation of A1 adenosine receptors. The antiplatelet and anti−inflammatory actions of adenosine appear to be mediated via the A2 adenosine receptor. In contrast, adenosine appears to be a pro-inflammatory mediator in asthma and chronic obstructive pulmonary disease (COPD).
Endogenous neurotransmitter. Cardioprotective effects may relate to activation of A1 adenosine receptors. The antiplatelet and anti−inflammatory actions of adenosine appear to be mediated via the A2 adenosine receptor. In contrast, adenosine appears to be a pro-inflammatory mediator in asthma and chronic obstructive pulmonary disease (COPD).
Tissue injury, ischemia and abnormal cell proliferation stimulate the release of adenosine. Adenosine helps in countering the effects of reduced oxygen and energy in organs such as heart, brain and skeletal muscles mediating vasodialation. It delivers its action through G protein-coupled receptors. Adenosine also reduces the release of neurotransmitters and cellular metabolism.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A9251PHR11381012123
form

powder

form

powder

form

-

form

solid

assay

≥99% (HPLC)

assay

≥99%

assay

-

assay

-

technique(s)

cell culture | mammalian: suitable

technique(s)

-

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

Quality Level

300

Quality Level

200

Quality Level

300

Quality Level

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

solubility

1 M NH4OH: 50 mg/mL, clear, colorless

solubility

-

solubility

-

solubility

-


Still not finding the right product?

Explore all of our products under Adenosine


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters