Skip to Content
Merck
All Photos(2)

Key Documents

C4438

Sigma-Aldrich

2-Chloro-2′-deoxyadenosine

antileukemic

Synonym(s):

CdA, Cladribine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12ClN5O3
CAS Number:
Molecular Weight:
285.69
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Quality Level

Assay

≥98% (HPLC)

form

powder or crystals

storage temp.

2-8°C

SMILES string

Nc1nc(Cl)nc2n(cnc12)[C@H]3C[C@H](O)[C@@H](CO)O3

InChI

1S/C10H12ClN5O3/c11-10-14-8(12)7-9(15-10)16(3-13-7)6-1-4(18)5(2-17)19-6/h3-6,17-18H,1-2H2,(H2,12,14,15)/t4-,5+,6+/m0/s1

InChI key

PTOAARAWEBMLNO-KVQBGUIXSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

2-Chloro-2′-deoxyadenosine (2-CdA) is a chlorinated purine nucleoside with activity against lymphoproliferative disorders, such as hairy cell leukemia (HCL) and multiple myeloma (MM). 2-CdA resists ADA degradation and is phosphorylated to CdATP in lymphocytes. CdATP incorporation into DNA induces strand breaks and the activation of apoptosis. 2-CdA may also be used in studies involving the inhibition of DNA polymerase(s).
Cladribiane, like fludarabine, is a prodrug that is must be phosphorylated intracellularly to the monophosphate by the nuclear/cytosol enzyme deoxycytidine kinase (dCK) and possibly by the mitochondrial enzyme deoxyguanosine kinase (dGK).
Cladribiane, like fludarabine, is a prodrug that is must be phosphorylated intracellularly to the monophosphate by the nuclear/cytosol enzyme deoxycytidine kinase (dCK) and possibly by the mitochondrial enzyme deoxyguanosine kinase (dGK). Clinically used for treatment of hairy cell leukemia, chronic lymphocytic leukemia and other indolent leukemias.

Biochem/physiol Actions

Deoxyadenosine analog resistant to adenosine deaminase; antileukemic with immunosuppressive activity

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Muta. 2 - Repr. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Gandhi et al.
Blood, 87(1), 256-264 (1996-01-01)
The effectiveness of arabinosylcytosine (ara-C) for the treatment of acute myelogenous leukemia (AML) depends on the formation of its active metabolite, the triphosphate of ara-C (ara-CTP). Using biochemical modulation strategies to increase the accumulation of ara-CTP in leukemia blasts, a
Tadeusz Robak et al.
Molecules (Basel, Switzerland), 14(3), 1183-1226 (2009-03-28)
For the past few years more and more new cytotoxic agents active in the treatment of hematological malignancies have been synthesized and become available for either in vitro studies or clinical trials. Among them the class of antineoplastic drugs belonging
D A Carson et al.
Proceedings of the National Academy of Sciences of the United States of America, 81(7), 2232-2236 (1984-04-01)
The adenosine deaminase-resistant purine deoxynucleoside 2-chloro-2'-deoxyadenosine (CdA) is markedly toxic in vitro to nondividing and proliferating normal human lymphocytes and to many leukemia cell specimens. The CdA is also effective against mouse L1210 leukemia in vivo. The present investigations have
H M Bryson et al.
Drugs, 46(5), 872-894 (1993-11-01)
Cladribine (2-chloro-2'-deoxyadenosine) is an adenosine deaminase-resistant analogue of deoxyadenosine. In the treatment of hairy cell leukaemia, cladribine has demonstrated excellent efficacy (complete response in 33 to 92% of patients) in noncomparative studies. Cladribine appears to compare favourably with other systemic
L Bastin-Coyette et al.
Biochemical pharmacology, 81(5), 586-593 (2010-12-21)
Nucleoside analogs (NAs) represent an important class of anticancer agents that induce cell death after conversion to triphosphate derivatives. One of their most important mechanisms of action is the activation of p53, leading to apoptosis through the intrinsic pathway. Classically

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service