Skip to Content
Merck
All Photos(1)

Key Documents

C2806

Sigma-Aldrich

Calcitonin Gene Related Peptide Fragment 8-37 human

≥97% (HPLC), powder

Synonym(s):

α-CGRP 8-37, FBHuman

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C139H230N44O38
CAS Number:
Molecular Weight:
3125.59
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥97% (HPLC)

form

powder

composition

Peptide content, ~70%

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)CNC(=O)[C@H](C)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](Cc1c[nH]cn1)NC(=O)[C@@H](NC(=O)[C@@H](N)C(C)C)[C@@H](C)O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CO)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CO)C(=O)NCC(=O)NCC(=O)N[C@@H](C(C)C)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](Cc2ccccc2)C(=O)N[C@@H](C(C)C)C(=O)N3CCC[C@H]3C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](C(C)C)C(=O)NCC(=O)N[C@@H](CO)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](C)C(=O)N[C@@H](Cc4ccccc4)C(N)=O

InChI

1S/C139H230N44O38/c1-66(2)48-86(161-102(193)60-155-113(197)74(17)159-120(204)87(49-67(3)4)168-118(202)83(40-31-45-152-138(147)148)163-123(207)90(53-80-57-151-65-158-80)173-136(220)111(77(20)188)182-131(215)105(145)69(7)8)121(205)169-88(50-68(5)6)122(206)176-96(64-186)129(213)165-84(41-32-46-153-139(149)150)119(203)175-94(62-184)115(199)156-58-101(192)154-59-104(195)177-107(71(11)12)134(218)179-108(72(13)14)133(217)166-82(39-28-30-44-141)117(201)171-91(54-98(142)189)125(209)172-92(55-99(143)190)124(208)170-89(52-79-36-25-22-26-37-79)126(210)180-109(73(15)16)137(221)183-47-33-42-97(183)130(214)181-110(76(19)187)135(219)174-93(56-100(144)191)127(211)178-106(70(9)10)132(216)157-61-103(194)162-95(63-185)128(212)164-81(38-27-29-43-140)116(200)160-75(18)114(198)167-85(112(146)196)51-78-34-23-21-24-35-78/h21-26,34-37,57,65-77,81-97,105-111,184-188H,27-33,38-56,58-64,140-141,145H2,1-20H3,(H2,142,189)(H2,143,190)(H2,144,191)(H2,146,196)(H,151,158)(H,154,192)(H,155,197)(H,156,199)(H,157,216)(H,159,204)(H,160,200)(H,161,193)(H,162,194)(H,163,207)(H,164,212)(H,165,213)(H,166,217)(H,167,198)(H,168,202)(H,169,205)(H,170,208)(H,171,201)(H,172,209)(H,173,220)(H,174,219)(H,175,203)(H,176,206)(H,177,195)(H,178,211)(H,179,218)(H,180,210)(H,181,214)(H,182,215)(H4,147,148,152)(H4,149,150,153)/t74-,75-,76+,77+,81-,82-,83-,84-,85-,86-,87-,88-,89-,90-,91-,92-,93-,94-,95-,96-,97-,105-,106-,107-,108-,109-,110-,111-/m0/s1

InChI key

NDACAFBDTQIYCQ-YVQXRMNASA-N

Gene Information

human ... CALCA(796)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Val-Thr-His-Arg-Leu-Ala-Gly-Leu-Leu-Ser-Arg-Ser-Gly-Gly-Val-Val-Lys-Asn-Asn-Phe-Val-Pro-Thr-Asn-Val-Gly-Ser-Lys-Ala-Phe-NH2

General description

Calcitonin-related peptide α (CALCA) is a 37-amino-acid vasoactive neuropeptide. It is scattered in the central and peripheral nervous systems. The calcitonin/calcitonin gene-related peptide gene complex codes for a small family of peptides: calcitonin, katacalcin and CALCA. This vasodilator is potent than other vasodilators, such as adenosine, substance P or acetylcholine. CALCA gene is located on human chromosome 11p15.2-p15.1. Procalcitonin (PCT) is a calcitonin (CT) precursor.

Application

Calcitonin Gene Related Peptide Fragment 8-37 human has been used in the administration of drugs. It has been used to test the hypothesis that calcitonin gene‐related peptide (CGRP) plays a role in the fetal cardiovascular and metabolic defense responses to acute hypoxaemia by affecting sympathetic outflow.

Biochem/physiol Actions

Selective competitive antagonist at CGRP receptors, but not at calcitonin receptors.
Calcitonin gene-related peptide (CALCA) serves as a proangiogenic growth factor in the development of human placenta. Mutations of CALCA gene is associated with Parkinson′s disease, ovarian cancer and essential hypertension. CALCA induce vascular endothelial cell proliferation, migration and capillary-like tube formation.

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calcitonin gene-related peptide (CALCA) is a proangiogenic growth factor in the human placental development
Dong Y L, et al.
Biology of Reproduction, 76(5), 892-899 (2007)
Regulation of pulpal microcirculation by calcitonin gene-related peptide
Kim S K, et al.
The Journal of Korean Academy of Prosthodontics, 30(6), 470-476 (2005)
Andressa Perin-Martins et al.
Journal of the peripheral nervous system : JPNS, 18(1), 62-74 (2013-03-26)
The aim of this study was to investigate the mechanisms that contribute to hyperalgesia and edema induced by TRPA1 activation. The injection of allyl isothiocyanate (AITC, 50, 100, or 300 µg/paw) into the rat's hind paw induced dose and time-dependent hyperalgesia
K J Escott et al.
British journal of pharmacology, 110(2), 772-776 (1993-10-01)
1. The effect of the calcitonin gene-related peptide antagonist (CGRP8-37, 400 nmol kg-1, i.v.) on the increased blood flow induced by calcitonin gene related peptide (CGRP), vasodilator prostaglandins, and topical capsaicin was measured with a laser Doppler blood flow meter
Association Between the Calcitonin-Related Peptide alpha(CALCA) Gene and Essential Hypertension in Japanese Subjects
Morita A, et al.
American Journal of Hypertension, 20(5), 527-532 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service