Skip to Content
Merck
All Photos(2)

Key Documents

C277

Sigma-Aldrich

Chloro-IB-MECA

solid, ≥98% (HPLC)

Synonym(s):

2-Chloro-N6-(3-iodobenzyl)-adenosine-5′-N-methyluronamide, Cl-IB-MECA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18ClIN6O4
CAS Number:
Molecular Weight:
544.73
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

color

white

solubility

DMSO: >20 mg/mL

SMILES string

CNC(=O)[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(NCc4cccc(I)c4)nc(Cl)nc23

InChI

1S/C18H18ClIN6O4/c1-21-16(29)13-11(27)12(28)17(30-13)26-7-23-10-14(24-18(19)25-15(10)26)22-6-8-3-2-4-9(20)5-8/h2-5,7,11-13,17,27-28H,6H2,1H3,(H,21,29)(H,22,24,25)/t11-,12+,13-,17+/m0/s1

InChI key

IPSYPUKKXMNCNQ-PFHKOEEOSA-N

Biochem/physiol Actions

2-chloro-N(6)-(3-iodobenzyl)adenosine-5′-N-methylcarboxamide (Cl-IB-MECA) is a adenosine receptor. It protects against myocardial ischemia/reperfusion injury in mice.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The A3 adenosine receptor agonist 2-Cl-IB-MECA facilitates epileptiform discharges in the CA3 area of immature rat hippocampal slices
Laudadio MA, et al.
Epilepsy Research, 59(2), 83-94 (2004)
K Stambaugh et al.
The American journal of physiology, 273(1 Pt 2), H501-H505 (1997-07-01)
The possible cardioprotective roles of adenosine A1 and A3 receptors were investigated in a cardiac myocyte model of injury. The adenosine A3 receptor is a novel cardiac receptor capable of mediating potentially important cardioprotective functions. Prolonged hypoxia with glucose deprivation
Cl-IB-MECA [2-Chloro-N6-(3-iodobenzyl) adenosine-5?-N-methylcarboxamide] Reduces Ischemia/Reperfusion Injury in Mice by Activating the A Adenosine Receptor
Ge ZD, et al.
Journal of Pharmacology and Experimental Therapeutics, 319(3), 1200-1210 (2006)
Y Yao et al.
Biochemical and biophysical research communications, 232(2), 317-322 (1997-03-17)
The effects of novel, selective adenosine (ADO) A3 receptor antagonists of diverse structure on cells of the human HL-60 leukemia and U-937 lymphoma cell lines were examined. Both 3-ethyl 5-benzyl 2-methyl-6-phenyl-4-phenylethynyl-1,4-(+/-)-dihydropyridine-3, 5-dicarboxylate (MRS 1191, 0.5 microM) and 6-carboxy-methyl-5, 9-dihydro-9-methyl-2-phenyl-[1,2,4]-triazolo [5,1-a][2,7]naphthyridine
Melanie Wurm et al.
Molecular cancer therapeutics, 20(11), 2250-2261 (2021-09-06)
Despite some impressive clinical results with immune checkpoint inhibitors, the majority of patients with cancer do not respond to these agents, in part due to immunosuppressive mechanisms in the tumor microenvironment. High levels of adenosine in tumors can suppress immune

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service