C2306
Anti-Rabbit IgG (whole molecule), F(ab′)2 fragment–Cy3 antibody produced in sheep
affinity isolated antibody, buffered aqueous solution
Synonym(s):
Anti-Rabbit IgG Cy3, Cy3 Anti-Rabbit IgG
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
sheep
conjugate
CY3 conjugate
antibody form
affinity isolated antibody
antibody product type
secondary antibodies
clone
polyclonal
form
buffered aqueous solution
species reactivity
rabbit
technique(s)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
unmodified
Related Categories
General description
Anti-Rabbit IgG (whole molecule), F(ab′)2 fragment–Cy3 antibody produced in sheep binds to all rabbit Igs and is useful when trying to avoid background staining due to the presence of Fc receptors.
The monomeric structure of immunoglobulin G (IgG) consists of two identical heavy chains and two identical light chains with molecular weight of 50kDa and 25kDa, respectively. The primary structure of this antibody also contains disulfide bonds involved in linking the two heavy chains, linking the heavy and light chains and resides inside the chains. IgG is further subdivided into four classes namely, IgG1, IgG2, IgG3, and IgG4 with different heavy chains, named γ1, γ2, γ3, and γ4, respectively. Limited digestion using papain cleaves the antibody into three fragments, two of which are identical and contain the antigen-binding activity (Fab fragments).
Application
Anti-Rabbit IgG (whole molecule), F(ab′)2 fragment-Cy3 antibody produced in sheep may be used for immunohistochemistry at a dilution of 1:100 and for immunocytochemistry.
Biochem/physiol Actions
IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections. The coupling of Cy3 to Anti-Rabbit IgG (whole molecule), F(ab′)2 fragment antibody allows for the visualization of proteins by fluorescent microscopy.
Physical form
Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide
Legal Information
Cy is distributed under license from Amersham Biosciences Limited.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Diana L Williams et al.
Neuropharmacology, 131, 83-95 (2017-12-10)
Glucagon-like peptide-1 (GLP-1) injected into the brain reduces food intake. Similarly, activation of preproglucagon (PPG) cells in the hindbrain which synthesize GLP-1, reduces food intake. However, it is far from clear whether this happens because of satiety, nausea, reduced reward
Tayaramma Thatava et al.
Stem cells (Dayton, Ohio), 24(12), 2858-2867 (2006-09-23)
Type 1 diabetes is caused by the destruction of pancreatic beta-cells by T cells of the immune system. Islet transplantation is a promising therapy for diabetes mellitus. Bone marrow stem cells (BMSC) have the capacity to differentiate into various cell
Yuan Wang et al.
International journal of molecular medicine, 40(2), 474-482 (2017-06-29)
The pathogenesis of Japanese encephalitis virus (JEV) is complex and unclearly defined, and in particular, the effects of the JEV receptor (JEVR) on diverse susceptible cells are elusive. In contrast to previous studies investigating JEVR in rodent or mosquito cells, in this
Jessica E Poort et al.
Human molecular genetics, 25(17), 3768-3783 (2016-08-06)
Spinal bulbar muscular atrophy (SBMA) is a progressive, late onset neuromuscular disease causing motor dysfunction in men. While the morphology of the neuromuscular junction (NMJ) is typically affected by neuromuscular disease, whether NMJs in SBMA are similarly affected by disease
Xinli Shi et al.
Oncotarget, 8(28), 45981-45993 (2017-05-21)
Dihydroartemisinin is an effective antimalarial agent with multiple biological activities. In the present investigation, we elucidated its therapeutic potential and working mechanism on human tongue squamous cell carcinoma (TSCC). It was demonstrated that dihydroartemisinin could significantly inhibit cell growth in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service