Skip to Content
Merck
All Photos(1)

Key Documents

C1017

Sigma-Aldrich

Citrinin

from Penicillium citrinum, ≥98% (HPLC), powder, mycotoxin

Synonym(s):

(3R,4S)-8-Hydroxy-3,4,5-Trimethyl-6-Oxo-4,6-Dihydro-3H-Isochromene-7-Carboxylic Acid, 4,6-Dihydro-8-hydroxy-3,4,5-trimethyl-6-oxo-3H-2-benzopyran-7-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H14O5
CAS Number:
Molecular Weight:
250.25
Beilstein:
5282243
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Citrinin, from Penicillium citrinum, ≥98% (HPLC)

biological source

Penicillium citrinum

Quality Level

Assay

≥98% (HPLC)

form

powder

storage temp.

2-8°C

SMILES string

C[C@H]1OC=C2C(O)=C(C(O)=O)C(=O)C(C)=C2[C@@H]1C

InChI

1S/C13H14O5/c1-5-7(3)18-4-8-9(5)6(2)11(14)10(12(8)15)13(16)17/h4-5,7,15H,1-3H3,(H,16,17)/t5-,7-/m1/s1

InChI key

CQIUKKVOEOPUDV-IYSWYEEDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Citrinin is a mycotoxin from Penicillium citrinin that comprises a quinone methide component.

Application

Citrinin has been used as:
  • a standard for calibration curve preparation for citrinin quantification Monascus purpureus ATCC 36928
  • an internal standard in thin layer chromatography (TLC) for characterizing the citrinin from Aspergillus and Penicillium strains
  • standard to characterize citrinin for rotten apples TLC

Biochem/physiol Actions

Citrinin is a predominant contaminant in agriculture supplies and food. It exhibits genotoxic, embryotoxic, and teratogenic properties. It is hepatonephrotoxin and a potent mycotoxin in rice grains.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liliana J G Silva et al.
Foods (Basel, Switzerland), 10(1) (2020-12-31)
Citrinin (CIT) deserves attention due to its known toxic effects in mammalian species and its widespread occurrence in food commodities, often along with ochratoxin A, another nephrotoxic mycotoxin. Human exposure, a key element in assessing risk related to food contaminants
M R Bragulat et al.
Letters in applied microbiology, 47(4), 286-289 (2009-02-26)
To assess the ability of fungi isolated from grapes to produce patulin and citrinin. A total of 446 Aspergillus isolates belonging to 20 species and 101 Penicillium isolates were inoculated in Czapek yeast extract agar and yeast extract sucrose agar
Effect of dissolved oxygen concentration on red pigment and citrinin production by Monascus purpureus ATCC 36928
Brazilian Journal of Chemical Engineering, 25, 247-253 (2008)
M L Martins et al.
Food additives and contaminants, 19(6), 568-574 (2002-06-04)
Patulin and citrinin are mycotoxins produced by certain fungi mainly belonging to Penicillium and Aspergillus and may be detected in mouldy fruits and fruit products. The data presented here refer to the simultaneous occurrence of patulin and citrinin in 351
Dubravka Flajs et al.
Arhiv za higijenu rada i toksikologiju, 60(4), 457-464 (2010-01-12)
Citrinin (CTN) is a nephrotoxic mycotoxin produced by several fungal strains belonging to the genera Penicillium, Aspergillus, and Monascus. It contaminates various commodities of plant origin, cereals in particular, and is usually found together with another nephrotoxic mycotoxin, ochratoxin A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service