Skip to Content
Merck
All Photos(3)

Documents

B4894

Sigma-Aldrich

Benzyl 2-acetamido-2-deoxy-α-D-galactopyranoside hydrate

O-glycosylation inhibitor, ≥97% (TLC)

Synonym(s):

Benzyl-α-GalNAc, α-D-GalNAc-1→OCH2Ph, Benzyl N-acetyl-α-D-galactosaminide, GalNAc α-O-benzyl, GalNAc-O-bn

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21NO6 · xH2O
CAS Number:
Molecular Weight:
311.33 (anhydrous basis)
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥97% (TLC)

form

powder

color

white to off-white

mp

204-208  °C

solubility

methanol: soluble 10 mg/mL, clear

storage temp.

−20°C

SMILES string

CC(N[C@H]([C@H]([C@H]1O)O)[C@H](O[C@@H]1CO)OCC2=CC=CC=C2)=O

InChI

InChI=1S/C15H21NO6/c1-9(18)16-12-14(20)13(19)11(7-17)22-15(12)21-8-10-5-3-2-4-6-10/h2-6,11-15,17,19-20H,7-8H2,1H3,(H,16,18)/t11-,12-,13+,14-,15+/m1/s1

InChI key

SKOZFDIGKDPQBO-QMIVOQANSA-N

Biochem/physiol Actions

Benzyl-N-acetyl-α-galactosaminide inhibits glycosyltransferase incorporation of glucosamine into O-glycans. Suppresses mucin biosynthesis and inhibits MUC1 expression in breast cancer cell line MDF-7.

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S F Kuan et al.
The Journal of biological chemistry, 264(32), 19271-19277 (1989-11-15)
Specific inhibitors of the glycosylation of O-glycosidically linked glycoproteins have not previously been described. When tested for their effects on mucin glycosylation in a mucin-producing colon cancer cell line, LS174T, benzyl-, phenyl-, and p-nitrophenyl-N-acetyl-alpha-galactosaminide inhibited the formation of fully glycosylated
Allison K Rodgers et al.
Fertility and sterility, 95(2), 823-825 (2010-10-26)
The attachment of endometrial epithelial cells (EECs) and endometrial stromal cells (ESCs) to peritoneal mesothelial cells (PMCs) with and without inhibition of N- and O-linked glycosylation, the viability of EECs and ESCs, and the expression of CD44 surface density were
Ashish V Kalra et al.
European journal of cancer (Oxford, England : 1990), 45(1), 164-173 (2008-12-03)
Current treatments for pancreatic cancer have failed to effectively manage the disease, and hence, more effective treatment approaches are urgently needed. Studies suggest that mucin O-glycosylation limits the cytotoxic effect of fluorouracil (5-FU) against the growth of human pancreatic cancer
Mika Sumiyoshi et al.
Investigative ophthalmology & visual science, 49(1), 197-203 (2008-01-04)
Prolonged contact of opposite mucosal surfaces, which occurs on the ocular surface, oral cavity, reproductive tract, and gut, requires a specialized apical cell surface that prevents adhesion. The purpose of this study was to evaluate the contribution of mucin O-glycans
A V Kalra et al.
British journal of cancer, 97(7), 910-918 (2007-10-04)
Mucins are high molecular weight glycoproteins expressed on the apical surface of normal epithelial cells. In cancer disease mucins are overexpressed on the entire cellular surface. Overexpression of MUC1 mucin in pancreatic tumours has been correlated with poor patient survival.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service