Skip to Content
Merck
All Photos(1)

Key Documents

B4560

Sigma-Aldrich

6-Bnz-cAMP sodium salt

≥98% (HPLC)

Synonym(s):

N6-Benzoyladenosine-3′,5′-cyclic monophosphate sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H15N5O7PNa
CAS Number:
Molecular Weight:
455.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

packaging

vial of 10 μmol

storage condition

desiccated

color

white

solubility

H2O: freely soluble

shipped in

dry ice

storage temp.

−70°C

SMILES string

[Na+].O[C@@H]1[C@@H]2OP([O-])(=O)OC[C@H]2O[C@H]1n3cnc4c(NC(=O)c5ccccc5)ncnc34

InChI

1S/C17H16N5O7P.Na/c23-12-13-10(6-27-30(25,26)29-13)28-17(12)22-8-20-11-14(18-7-19-15(11)22)21-16(24)9-4-2-1-3-5-9;/h1-5,7-8,10,12-13,17,23H,6H2,(H,25,26)(H,18,19,21,24);/q;+1/p-1/t10-,12-,13-,17-;/m1./s1

InChI key

SPYGSKQRPXISIB-FKVBDRBCSA-M

Biochem/physiol Actions

6-Bnz-cAMP is a membrane permeable and selective cAMP-dependent protein kinase (PKA) activator. For preferential stimulation of cAK type I, a combination with the site B selective analog 8-HA-cAMP or 8-AHA-cAMP can be used.

Features and Benefits

This compound is featured on the PKA & PKG page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kevin W-H Lo et al.
Journal of tissue engineering and regenerative medicine, 6(1), 40-48 (2011-02-12)
Osteoblastic differentiation is an important landmark for bone formation, bone repair and regeneration; however, it is a very complex process controlled by different signalling mechanisms. Several groups have reported that the cyclic adenosine monophosphate (cAMP) signalling system is responsible for
H Kita et al.
Journal of immunology (Baltimore, Md. : 1950), 146(8), 2712-2718 (1991-04-15)
We have investigated the effects of cAMP on Ig-induced human eosinophil activation. Stimulation of human normodense eosinophils with IgG- or secretory IgA (sIgA)-coated Sepharose beads induced cellular degranulation, as measured by the release of the granule protein, eosinophil-derived neurotoxin (EDN).
R Bøe et al.
British journal of cancer, 72(5), 1151-1159 (1995-11-01)
8-Cl-cAMP and 8-NH2-cAMP induced MCF-7 cell death. The type(s) of cell death were studied in more detail and compared with the cell death type (apoptosis) induced by okadaic acid, an inhibitor of serine/threonine phosphatases. By morphological criteria dying cells showed
R A Steinberg et al.
The Journal of biological chemistry, 271(44), 27630-27636 (1996-11-01)
The guanidinium groups of conserved arginines in the two intrachain cAMP-binding sites of regulatory (R) subunit of cAMP-dependent protein kinase have been implicated in the allosteric interactions by which cAMP binding leads to kinase activation. We have investigated the functional
B M Hokland et al.
The Journal of biological chemistry, 268(34), 25343-25349 (1993-12-05)
The interaction of high density lipoprotein with its putative receptor stimulates translocation and efflux of intracellular sterols by a process involving activation of protein kinase C. This study shows that activation of cAMP-dependent protein kinase also stimulates efflux of intracellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service