Skip to Content
Merck

B2643

Biotinamidohexanoic acid N-hydroxysuccinimide ester

≥98% (TLC), powder

Synonym(s):

(+)-Biotinamidocaproate N-hydroxysuccinimidyl ester, N-(+)-Biotinyl-6-aminocaproic acid N-succinimidyl ester, N-(+)-Biotinyl-6-aminohexanoic acid NHS ester, N-Succinimidyl N-biotinyl-6-aminocaproate, Succinimidyl 6-(biotinamido)hexanoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H30N4O6S
CAS Number:
Molecular Weight:
454.54
UNSPSC Code:
12352203
NACRES:
NA.46
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
3577403
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (TLC)

form

powder

solubility

H2O: ≤2 mg/mL (with sonication), DMF: 50 mg/mL (Stable up to 2 months in dry DMF.)

storage temp.

−20°C

SMILES string

[H][C@]12CS[C@@H](CCCCC(=O)NCCCCCC(=O)ON3C(=O)CCC3=O)[C@@]1([H])NC(=O)N2

InChI

1S/C20H30N4O6S/c25-15(7-4-3-6-14-19-13(12-31-14)22-20(29)23-19)21-11-5-1-2-8-18(28)30-24-16(26)9-10-17(24)27/h13-14,19H,1-12H2,(H,21,25)(H2,22,23,29)/t13-,14-,19-/m0/s1

InChI key

UVGHPGOONBRLCX-NJSLBKSFSA-N

Application

Biotinamidohexanoic acid N-hydroxysuccinimide ester has been used in the biotinylation of antibodies.

Biochem/physiol Actions

Biotin is a label widely used in several biochemical applications and has a high affinity for avidin. A spacer chain is required to reduce steric hindrance while coupling biotin (a small molecule) to biopolymers (large molecules). Biotinamidohexanoic acid N-hydroxysuccinimide ester incorporates a 7-atom spacer with the N-succinimide leaving group. It can react with a primary amine in the pH range of 6.5-8.5. This biotinylation reagent incorporates an aminocaproyl spacer and thus can reduce the steric hindrance in binding avidin to some biotinylated compounds.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library