Skip to Content
Merck

B3295

Biotinamidohexanoyl-6-aminohexanoic acid N-hydroxysuccinimide ester

≥95% (TLC), powder

Synonym(s):

6-(Biotinamidocaproylamido)caproic acid N-hydroxysuccinimide ester, Biotin-X-X-NHS, N-Succinimidyl N-[6-(biotinylamino)caproyl]-6-aminocaproate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C26H41N5O7S
CAS Number:
Molecular Weight:
567.70
UNSPSC Code:
12352106
NACRES:
NC.07
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
8377428
Assay:
≥95% (TLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (TLC)

form

powder

solubility

DMF: 25 mg/mL

storage temp.

−20°C

SMILES string

[H][C@]12CS[C@@H](CCCCC(=O)NCCCCCC(=O)NCCCCCC(=O)ON3C(=O)CCC3=O)[C@@]1([H])NC(=O)N2

InChI

1S/C26H41N5O7S/c32-20(27-16-8-2-4-12-24(36)38-31-22(34)13-14-23(31)35)10-3-1-7-15-28-21(33)11-6-5-9-19-25-18(17-39-19)29-26(37)30-25/h18-19,25H,1-17H2,(H,27,32)(H,28,33)(H2,29,30,37)/t18-,19-,25-/m0/s1

InChI key

ATYCFNRXENKXSE-MHPIHPPYSA-N

Application

Biotinylation reagent incorporating an extended 14-atom spacer can reduce steric hindrance in binding avidin or streptavidin to some biotinylated compounds and increase sensitivity for detection. Typically coupled to primary amine in the pH range 6.5-8.5.
Biotinamidohexanoyl-6-aminohexanoic acid N-hydroxysuccinimide ester has been used in human IgM biotinylation.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library