Skip to Content
Merck
All Photos(1)

Key Documents

B134

Sigma-Aldrich

BMY 7378 dihydrochloride

≥98% (HPLC), solid

Synonym(s):

8-[2-[4-(2-Methoxyphenyl)-1-piperazinyl]ethyl]-8-azaspiro[4.5]decane-7,9-dione dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H31N3O3 · 2HCl
CAS Number:
Molecular Weight:
458.42
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

form

solid

color

white

solubility

H2O: >5 mg/mL

SMILES string

Cl.Cl.COc1ccccc1N2CCN(CC2)CCN3C(=O)CC4(CCCC4)CC3=O

InChI

1S/C22H31N3O3.2ClH/c1-28-19-7-3-2-6-18(19)24-13-10-23(11-14-24)12-15-25-20(26)16-22(17-21(25)27)8-4-5-9-22;;/h2-3,6-7H,4-5,8-17H2,1H3;2*1H

InChI key

NIBOMXUDFLRHRV-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

BMY 7378 dihydrochloride is a partial 5-HT1A serotonin receptor agonist and selective α1D-adrenoceptor antagonist.

Features and Benefits

This compound is featured on the α1-Adrenoceptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Legal Information

Manufactured and sold with the permission of Bristol Myers Squibb Corporation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Yang et al.
Naunyn-Schmiedeberg's archives of pharmacology, 355(4), 438-446 (1997-04-01)
We have used the alpha1D-adrenoceptor selective antagonist, BMY 7378, the alpha1D-selective agonists, adrenaline and phenylephrine, the alpha1A-selective antagonists, (+)-niguldipine, SB 216469 and WB4101, and the non-subtype-selective alpha1-adrenoceptor antagonist, nemonapride, to investigate the presence of alpha1D-adrenoceptors in rat tissues at the
M J Millan et al.
The Journal of pharmacology and experimental therapeutics, 268(1), 337-352 (1994-01-01)
The novel benzodioxopiperazines [4-(benzodioxan-5-yl)1-[2- (benzocyclobutane-1-yl)ethyl]piperazine] (S 14489), [4-(benzodioxan-5-yl)1-(indan-2-yl)piperazine)] (S 15535) and [4-(benzodioxan-5-yl)1-[2(indan-1-yl)ethyl]piperazine (S15931) competitively displaced the binding of [3H]-8-OH-DPAT at serotonin (5-HT)1A receptors with affinities (pKis) of 9.2, 8.8 and 8.9, respectively. These values compared favorably with those of the
BMY 7378, a buspirone analog with high affinity, selectivity and low intrinsic activity at the 5-HT1A receptor in rat and guinea pig hippocampal membranes.
F D Yocca et al.
European journal of pharmacology, 137(2-3), 293-294 (1987-06-04)
M Eltze et al.
European journal of pharmacology, 378(1), 69-83 (1999-09-09)
The affinity for functional alpha1-adrenoceptor subtypes of buspirone in comparison with its close structural analogs and selective alpha1D-adrenoceptor antagonists, BMY 7378 (8-[2-[4-(2-methoxyphenyl)-1-piperazinyl]ethyl]-8-azaspiro[4.5]dec ane-7,9-dione) and MDL 73005EF (8-[2-(1,4-benzodioxan-2-ylmethylamino)ethyl]-8-azaspiro+ ++[4.5]decane-7,9-dione), was determined, namely at subtype A in rat vas deferens and perfused
T Sharp et al.
European journal of pharmacology, 176(3), 331-340 (1990-02-13)
The present study examined the actions of the putative 5-HT1A antagonist BMY 7378 on central pre- and postsynaptic 5-HT1A function in the rat in vivo. Unlike the direct acting 5-HT1A agonist 8-hydroxy-2-(di-n-pro-pylamino)tetralin (8-OH-DPAT), BMY 7378 (0.25-5 mg/kg s.c.) did not

Articles

Review alpha 1 adrenoceptors as well as their agonists, antagonists, and tissue expression patterns. We suggest several modulators and alternatives for working with a-1 adrenoreceptors.

Review alpha 1 adrenoceptors as well as their agonists, antagonists, and tissue expression patterns. We suggest several modulators and alternatives for working with a-1 adrenoreceptors.

Review alpha 1 adrenoceptors as well as their agonists, antagonists, and tissue expression patterns. We suggest several modulators and alternatives for working with a-1 adrenoreceptors.

Review alpha 1 adrenoceptors as well as their agonists, antagonists, and tissue expression patterns. We suggest several modulators and alternatives for working with a-1 adrenoreceptors.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service