Skip to Content
Merck
All Photos(2)

Key Documents

AV43558

Sigma-Aldrich

Anti-PEMT antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-MGC2483, Anti-PEAMT, Anti-PEMPT, Anti-PEMT2, Anti-PNMT, Anti-Phosphatidylethanolamine N-methyltransferase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

26 kDa

species reactivity

rat, rabbit, human, horse, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PEMT(10400)

General description

The previously assigned protein identifier Q9BW86 has been merged into Q9UBM1. Full details can be found on the UniProt database.

Immunogen

Synthetic peptide directed towards the C terminal region of human PEMT

Biochem/physiol Actions

PEMT is an enzyme which converts phosphatidylethanolamine to phosphatidylcholine by sequential methylation in the liver. The protein localizes to the endoplasmic reticulum and mitochondria-associated membranes. The gene encodes PEMT protein is within the Smith-Magenis syndrome region on chromosome 17. This gene encodes an enzyme which converts phosphatidylethanolamine to phosphatidylcholine by sequential methylation in the liver. The protein localizes to the endoplasmic reticulum and mitochondria-associated membranes. The gene is within the Smith-Magenis syndrome region on chromosome 17. Alternate splicing of this gene results in three transcript variants encoding two different isoforms.

Sequence

Synthetic peptide located within the following region: GWAIMHASPTGLLLTVLVALTYIVALLYEEPFTAEIYRQKASGSHKRS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marit Espe et al.
The British journal of nutrition, 112(8), 1274-1285 (2014-09-10)
During the last few decades, plant protein ingredients such as soya proteins have replaced fishmeal in the diets of aquacultured species. This may affect the requirement and metabolism of methionine as soya contains less methionine compared with fishmeal. To assess
Alba Capelo-Diz et al.
Cell metabolism, 35(8), 1373-1389 (2023-08-02)
There has been an intense focus to uncover the molecular mechanisms by which fasting triggers the adaptive cellular responses in the major organs of the body. Here, we show that in mice, hepatic S-adenosylmethionine (SAMe)-the principal methyl donor-acts as a
Y F Zhou et al.
Journal of dairy science, 101(12), 11384-11395 (2018-10-15)
Insufficient supply of Met and choline (Chol) around parturition could compromise hepatic metabolism and milk protein synthesis in dairy cows. Mechanistic responses associated with supply of Met or Chol in primary liver cells enriched with hepatocytes (PHEP) from cows have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service