Skip to Content
Merck
All Photos(1)

Documents

L6785

Sigma-Aldrich

Lactacystin

≥90% (HPLC), powder, proteosome inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24N2O7S
CAS Number:
Molecular Weight:
376.43
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Lactacystin, ≥90% (HPLC)

Quality Level

Assay

≥90% (HPLC)

form

powder

potency

4 nM Ki (proteasome inhibitor)

solubility

water: 10 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

CC(C)[C@H](O)[C@]1(NC(=O)[C@H](C)[C@@H]1O)C(=O)SC[C@H](NC(C)=O)C(O)=O

InChI

1S/C15H24N2O7S/c1-6(2)10(19)15(11(20)7(3)12(21)17-15)14(24)25-5-9(13(22)23)16-8(4)18/h6-7,9-11,19-20H,5H2,1-4H3,(H,16,18)(H,17,21)(H,22,23)/t7-,9+,10+,11+,15-/m1/s1

InChI key

DAQAKHDKYAWHCG-RWTHQLGUSA-N

General description

Lactacystin is an antibiotic and a metabolite of Streptomyces spp.

Application

Lactacystin has been used:
  • as a proteasome inhibitor to inhibit protein degradation
  • to inhibit proteasomal activity of cells for live cell imaging
  • to block proteasomal proteolysis in human monocyte-derived dendritic cells (MoDCs) for 24 h
  • to provide unilateral injection to animals to induce nigrostriatal lesions

Biochem/physiol Actions

Lactacystin can block the development of cell cycle and stimulate differentiation in a murine neuroblastoma cell line. It can serve as a precursor for clasto-lactacystin β-lactone. Cell-permeable and irreversible proteasome inhibitor (Ki = 4nM). Inhibits NF-kB activation (IC50 = 10mM). Induces neurite outgrowth in neuro2A mouse neuroblastoma cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Differential regulation of MHC II and invariant chain expression during maturation of monocyte-derived dendritic cells
Landsverk O J B, et al.
Journal of Leukocyte Biology, 91(5), 729-737 (2012)
Mechanistic Studies on the Inactivation of the Proteasome by Lactacystin A CENTRAL ROLE FOR clasto-LACTACYSTIN beta-LACTONE
Dick L R, et al.
The Journal of Biological Chemistry, 271(13), 7273-7276 (1996)
Cannabinoid receptor-induced neurite outgrowth is mediated by Rap1 activation through Galphao/i-triggered proteasomal degradation of Rap1GAPII
Jordan J D, et al.
The Journal of Biological Chemistry, 280(12), 11413-11421 (2005)
The development and pharmacology of proteasome inhibitors for the management and treatment of cancer
Ruggeri B, et al.
Advances in Pharmacology, 57(13), 91-135 (2009)
Evolution of extra-nigral damage predicts behavioural deficits in a rat proteasome inhibitor model of Parkinson's disease
Vernon A C, et al.
PLoS ONE, 6(2), e17269-e17269 (2011)

Articles

Discover definitions and uses for irreversible inhibitors including the types of irreversible inhibitors: suicide inhibitors, heavy metal inhibitors, and time-dependent inhibitors.

Discover definitions and uses for irreversible inhibitors including the types of irreversible inhibitors: suicide inhibitors, heavy metal inhibitors, and time-dependent inhibitors.

Discover definitions and uses for irreversible inhibitors including the types of irreversible inhibitors: suicide inhibitors, heavy metal inhibitors, and time-dependent inhibitors.

Discover definitions and uses for irreversible inhibitors including the types of irreversible inhibitors: suicide inhibitors, heavy metal inhibitors, and time-dependent inhibitors.

See All

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service