Skip to Content
Merck

A8980

Acarbose

≥95% (HPLC)

Synonym(s):

4",6"-Dideoxy-4"-([1S]-[1,4,6/5]-4,5,6-trihydroxy-3-hydroxymethyl-2-yclohexenylamino)-maltotriose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C25H43NO18
CAS Number:
Molecular Weight:
645.60
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352201
EC Number:
260-030-7
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bacterial (Actinoplanes)

Quality Segment

assay

≥95% (HPLC)

form

powder

impurities

<9.4% water (Karl Fischer)

color

white to off-white

solubility

H2O (CO2 free): 50 mg/mL, clear, colorless

storage temp.

room temp

SMILES string

C[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O[C@@H]2CO)O[C@H]3[C@H](O)[C@@H](O)[C@H](O)O[C@@H]3CO)[C@H](O)[C@@H](O)[C@@H]1N[C@H]4C=C(CO)[C@@H](O)[C@H](O)[C@H]4O

InChI

1S/C25H43NO18/c1-6-11(26-8-2-7(3-27)12(30)15(33)13(8)31)14(32)19(37)24(40-6)43-22-10(5-29)42-25(20(38)17(22)35)44-21-9(4-28)41-23(39)18(36)16(21)34/h2,6,8-39H,3-5H2,1H3/t6-,8+,9-,10-,11-,12-,13+,14+,15+,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-/m1/s1

InChI key

XUFXOAAUWZOOIT-SXARVLRPSA-N

Gene Information

Application

Acarbose, a derivitized maltotriose antidiabetic drug, is a reversible α-glucosidase inhibitor used in medical studies to control the effects of Type II diabetes. Acarbose is often used as a reference to which other α-glucosidase inhibitors are compared.

Biochem/physiol Actions

Modified tetrasaccharide that acts as a reversible α -glucosidase inhibitor.

Other Notes

To gain a comprehensive understanding of our extensive range of Oligosaccharides for your research, we encourage you to visit our Carbohydrates Category page.


Still not finding the right product?

Explore all of our products under Acarbose


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library