![Acarbose European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/343/152/d24f1e70-e230-43d4-89ba-f098bd03eb6f/640/d24f1e70-e230-43d4-89ba-f098bd03eb6f.png)
Acarbose
![Acarbose European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/343/152/d24f1e70-e230-43d4-89ba-f098bd03eb6f/640/d24f1e70-e230-43d4-89ba-f098bd03eb6f.png)
Synonym(s):
4",6"-Dideoxy-4"-([1S]-[1,4,6/5]-4,5,6-trihydroxy-3-hydroxymethyl-2-yclohexenylamino)-maltotriose, Acarbose
Empirical Formula (Hill Notation):
C25H43NO18
CAS Number:
Molecular Weight:
645.60
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Analyte Suitability
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique