Skip to Content
Merck
All Photos(1)

Key Documents

A7155

Sigma-Aldrich

p-Aminobenzamidine–Agarose

saline suspension

Synonym(s):

p-ABA

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106514
NACRES:
NA.56

form

saline suspension

Quality Level

matrix

cross-linked 4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

amino to carboxyl of spacer

matrix spacer

7 atoms

capacity

≥10 mg/mL binding capacity (trypsin)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

p-Aminobenzamidine-Agarose (pABA) can be used as an affinity ligand to bind and remove serine proteases in addition to Ca2+-binding proteins. It has also been used in medical applications as a tool for the purification of non-protease proteins.

Physical form

Suspension in 0.5 M NaCl containing preservative

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

482.0 °F - closed cup

Flash Point(C)

250 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Anisowicz et al.
Molecular medicine (Cambridge, Mass.), 2(5), 624-636 (1996-09-01)
Using differential display (DD), we discovered a new member of the serine protease family of protein-cleaving enzymes, named protease M. The gene is most closely related by sequence to the kallikreins, to prostate-specific antigen (PSA), and to trypsin. The diagnostic
Andrew M Petros et al.
Journal of medicinal chemistry, 62(8), 4120-4130 (2019-04-02)
Apolipoprotein E is a 299-residue lipid carrier protein produced in both the liver and the brain. The protein has three major isoforms denoted apoE2, apoE3, and apoE4 which differ at positions 112 and 158 and which occur at different frequencies
J F Eccleston et al.
The Journal of biological chemistry, 268(36), 27012-27019 (1993-12-25)
Single turnover and equilibrium binding measurements on the interaction of Gly-12 and Pro-12 Ras.GTP with the catalytic domains of the GTPase-activating proteins, p120-GAP and neurofibromin, have been made utilizing fluorescent 2'(3')O-(N-methylanthraniloyl)-nucleotides. These have enabled the equilibrium dissociation constants (Kd) for
The distinctive cell division interactome of Neisseria gonorrhoeae.
Zou, et al.
BMC Microbiology, 17, 232-232 (2018)
Biochemical and molecular modeling analysis of the ability of two p-aminobenzamidine-based sorbents to selectively purify serine proteases (fibrinogenases) from snake venoms.
De-Simone, S.G., et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 822(1-2), 1-9 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service