Skip to Content
Merck

A2411

Amphotericin B from Streptomyces sp.

solid, suitable for cell culture, BioReagent

Synonym(s):

Fungizone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C47H73NO17
CAS Number:
Molecular Weight:
924.08
NACRES:
NA.76
PubChem Substance ID:
UNSPSC Code:
12352207
EC Number:
215-742-2
MDL number:
Beilstein/REAXYS Number:
78342
eCl@ss:
34010400
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Amphotericin B from Streptomyces sp., BioReagent, suitable for cell culture, ~80% (HPLC)

biological source

Streptomyces sp.

Quality Segment

product line

BioReagent

assay

~80% (HPLC)

form

solid

potency

>750.00 μg per mg (Dry basis)

technique(s)

cell culture | mammalian: suitable

color

yellow to yellow-orange

solubility

H2O: slightly soluble

antibiotic activity spectrum

fungi, yeast

mode of action

cell membrane | interferes

storage temp.

2-8°C

SMILES string

C[C@H]1O[C@@H](O[C@@H]2C[C@@H]3O[C@](O)(C[C@@H](O)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)CC(=O)O[C@@H](C)[C@H](C)[C@H](O)[C@@H](C)\C=C\C=C\C=C\C=C\C=C\C=C\C=C\2)C[C@H](O)[C@H]3C(O)=O)[C@@H](O)[C@@H](N)[C@@H]1O

InChI

1S/C47H73NO17/c1-27-17-15-13-11-9-7-5-6-8-10-12-14-16-18-34(64-46-44(58)41(48)43(57)30(4)63-46)24-38-40(45(59)60)37(54)26-47(61,65-38)25-33(51)22-36(53)35(52)20-19-31(49)21-32(50)23-39(55)62-29(3)28(2)42(27)56/h5-18,27-38,40-44,46,49-54,56-58,61H,19-26,48H2,1-4H3,(H,59,60)/b6-5+,9-7+,10-8+,13-11+,14-12+,17-15+,18-16+/t27-,28-,29-,30+,31+,32+,33-,34-,35+,36+,37-,38-,40+,41-,42+,43+,44-,46-,47+/m0/s1

InChI key

APKFDSVGJQXUKY-INPOYWNPSA-N

General description

Chemical structure: polyene

Application

Amphotericin B is a polyene antifungal antibiotic from Streptomyce sp. It has a high affinity for sterols, especially ergosterols, of both fungal and bacterial cell membranes. Normal usage in maintenance of antifungal cell cultures is 2.5 mg/L with penicillin and streptomycin used in the medium.

Biochem/physiol Actions

Amphotericin B binds to sterols, forming pores in the membrane, and causing small molecules to leak out. The product is most effective against fungi and yeast.

Preparation Note

Amphotericin B is insoluble in water at pH 6 to 7, but soluble in water at pH 2 or 11. It is soluble in DMSO (30-40 mg/mL) and in dimethylformamide (2-4 mg/mL). Aqueous solutions cannot be sterile filtered due to poor solubility.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Product is sensitive to light and moisture.

Disclaimer

Amphotericin B remains active for 3 days in culture at 37°C. At -20°C, and with protection from air and light, the product remains active for 5 years.


Still not finding the right product?


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

STOT RE 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library