Skip to Content
Merck

Skip To

A6779

Adenosine 5′-triphosphate, periodate oxidized sodium salt

≥97%, bacterial (corynebacterium sp), powder

Synonym(s):

oATP, Adenosine 5′-triphosphate-2′,3′-dialdehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10H14N5O13P3
CAS Number:
Molecular Weight:
505.17
UNSPSC Code:
41106305
eCl@ss:
32160414
PubChem Substance ID:
NACRES:
NA.51
MDL number:
Assay:
≥97%
Biological source:
bacterial (corynebacterium sp)
Form:
powder
Storage temp.:
−20°C
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Adenosine 5′-triphosphate, periodate oxidized sodium salt, ≥97%

biological source

bacterial (corynebacterium sp)

Quality Segment

assay

≥97%

form

powder

storage temp.

−20°C

SMILES string

[Na].Nc1ncnc2n(cnc12)[C@H](O[C@H](COP(O)(=O)OP(O)(=O)OP(O)(O)=O)C=O)C=O

InChI

1S/C10H14N5O13P3.Na.H/c11-9-8-10(13-4-12-9)15(5-14-8)7(2-17)26-6(1-16)3-25-30(21,22)28-31(23,24)27-29(18,19)20;;/h1-2,4-7H,3H2,(H,21,22)(H,23,24)(H2,11,12,13)(H2,18,19,20);;/t6-,7+;;/m0../s1

InChI key

IHQATUPGVKMGLT-AUCRBCQYSA-N

General description

Adenosine 5′-triphosphate 2′,3′ dialdehyde is a derivative of dialdehyde.

Application

Adenosine 5′-triphosphate, periodate oxidized sodium salt has been used:
  • as an antagonist of the purinergic receptor, P2 × 7 to inhibit P2 × 7 pathway
  • as a nonspecific agonist to assess the activity of P2×7-R by the mobilization of Ca2+
  • to evaluate the physiological significance of elevated intracellular ATP levels
  • increased P2rx7 expression for osteoclastogenesis in Flcn-deficient cells

Biochem/physiol Actions

Adenosine 5′-triphosphate 2′,3′ dialdehyde is considered as an inhibitor of P2 × 7 receptor.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A6904A5638A9376
biological source

bacterial (corynebacterium sp)

biological source

-

biological source

-

biological source

synthetic (organic)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

powder

form

solid

form

powder

form

powder

assay

≥97%

assay

90-95%

assay

≥95%

assay

≥93%

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters