Skip to Content
Merck

A6904

Adenosine 5′-diphosphate, periodate oxidized sodium salt

90-95%

Synonym(s):

oADP, Adenosine 5′-diphosphate-2′,3′-dialdehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H13N5O10P2
CAS Number:
Molecular Weight:
425.19
NACRES:
NA.51
PubChem Substance ID:
UNSPSC Code:
41106305
MDL number:
Assay:
90-95%
Form:
solid
Storage temp.:
−20°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

90-95%

form

solid

storage temp.

−20°C

SMILES string

[Na].Nc1ncnc2n(cnc12)[C@H](O[C@H](COP(O)(=O)OP(O)(O)=O)C=O)C=O

InChI

1S/C10H13N5O10P2.Na.H/c11-9-8-10(13-4-12-9)15(5-14-8)7(2-17)24-6(1-16)3-23-27(21,22)25-26(18,19)20;;/h1-2,4-7H,3H2,(H,21,22)(H2,11,12,13)(H2,18,19,20);;/t6-,7+;;/m0../s1

InChI key

VWUMWPCYVOHNRD-AUCRBCQYSA-N

Application

Adenosine 5′-diphosphate-2′,3′-dialdehyde (oADP) is used to stimulate the permeability transition in mitochondria. oADP and oATP modify the duration and intensity of firefly luciferase-containing reactions. oADP has been used as an affinity label for the NAD+ binding site of recombinant Candida boidinii formate dehydrogenase (FDH) and to inactivate muscle pyruvate kinase.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library