Skip to Content
Merck
All Photos(5)

Key Documents

A6680

Sigma-Aldrich

N-Acetylneuraminic Acid Aldolase from microorganisms

lyophilized powder, ≥20 units/mg protein (biuret)

Synonym(s):

N-Acetylneuraminate Pyruvate Lyase, N-Acetylneuraminic Acid Lyase, NANA Aldolase, Sialic Aldolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
Beilstein:
2697172
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32
Pricing and availability is not currently available.

form

lyophilized powder

specific activity

≥20 units/mg protein (biuret)

mol wt

~98 kDa

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

This enzyme is useful for enzymatic determination of N-acetylneuraminic acid and sialic acid when
coupled with the related enzymes in clinical analysis.
For industrial use, this enzyme is useful for enzymatic synthesis of sialic acid.
Used in the Sialic Acid Quantification Kit, SIALIC-Q

Physical properties

Isoelectric point: 4.6 ± 0.1
Michaelis constant: 2.5 x 10‾3M (N-Acetylneuraminic acid)
Structure: 3 subunits (approx. 35,000) per mol of enzyme
Inhibitors: p-Chloromercuribenzoate, sodium dodecyl sulfact, Hg++, Ag+
Optimum pH: 7.5– 8.0
Optimum temp: 70°C
pH Stability: pH 6.0–9.0 (10°C, 25hr)
Thermal stability: Below 65°C (pH 7.5, 30 min)

Unit Definition

One unit will release 1.0 μmole of pyruvate from NANA per min at pH 7.7 at 37 °C.

Physical form

Lyophilized powder containing mannitol and EDTA

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Varun Bhaskar et al.
Proteins, 79(4), 1132-1142 (2011-02-05)
YagE is a 33 kDa prophage protein encoded by CP4-6 prophage element in Escherichia coli K12 genome. Here, we report the structures of YagE complexes with pyruvate (PDB Id 3N2X) and KDGal (2-keto-3-deoxy galactonate) (PDB Id 3NEV) at 2.2A resolution.
Yanhong Li et al.
Applied microbiology and biotechnology, 79(6), 963-970 (2008-06-04)
Sialic acid aldolases or N-acetylneuraminate lyases (NanAs) catalyze the reversible aldol cleavage of N-acetylneuraminic acid (Neu5Ac) to form pyruvate and N-acetyl-D: -mannosamine (ManNAc). A capillary electrophoresis assay was developed to directly characterize the activities of NanAs in both Neu5Ac cleavage
Hee Gon Jeong et al.
Infection and immunity, 77(8), 3209-3217 (2009-06-03)
N-acetylneuraminic acid (Neu5Ac, sialic acid) could provide a good substrate for enteropathogenic bacteria in the intestine, when the bacteria invade and colonize in human gut. In order to analyze the role of Neu5Ac catabolism in Vibrio vulnificus pathogenesis, a mutant
Jozef Nahálka et al.
Journal of biotechnology, 134(1-2), 146-153 (2008-03-04)
The propensity of a recombinant protein produced in bacteria to aggregate has been assumed to be unpredictable, and inclusion bodies have been thought of as wasted cell material. However, a target protein can be purposely driven to inclusion bodies, which
Wen-liu Yang et al.
Zhejiang da xue xue bao. Yi xue ban = Journal of Zhejiang University. Medical sciences, 39(1), 57-63 (2010-02-23)
To obtain the Escherichia coli strains expressing N-Acetyl-D-neuraminic acid aldolase (Neu5Ac aldolase). The gene (nanA) coding Neu5Ac aldolase was cloned from Escherichia coli C600, and the recombinant plasmid was sequenced and expressed in Escherichia coli. Sequencing data revealed that the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service