Skip to Content
Merck

Skip To

A4906

(+)-Abscisic acid

≥98% (HPLC), powder, growth regulator

Synonym(s):

(S)-5-(1-Hydroxy-2,6,6-trimethyl-4-oxo-2-cyclohexen-1-yl)-3-methyl-(2Z,4E)-pentadienoic acid, ABA, Dormin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C15H20O4
CAS Number:
Molecular Weight:
264.32
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352106
EC Number:
244-319-5
MDL number:
Beilstein/REAXYS Number:
2698956
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

(+)-Abscisic acid, ≥98% (HPLC)

Quality Segment

assay

≥98% (HPLC)

form

powder

application(s)

agriculture

storage temp.

−20°C

SMILES string

CC(\C=C\[C@@]1(O)C(C)=CC(=O)CC1(C)C)=C\C(O)=O

InChI

1S/C15H20O4/c1-10(7-13(17)18)5-6-15(19)11(2)8-12(16)9-14(15,3)4/h5-8,19H,9H2,1-4H3,(H,17,18)/b6-5+,10-7-/t15-/m1/s1

InChI key

JLIDBLDQVAYHNE-YKALOCIXSA-N

Application

(+)-Abscisic acid has been used:
  • as a supplement for the germination of Arabidopsis thaliana seeds
  • as a component of buffered nodulation medium to check its effects on lateral root density (LRD) in leguminous and non-leguminous plants
  • as a standard to study its effects on root meristem defects in Medicago truncatula latd mutant

Biochem/physiol Actions

Plant hormone and growth regulator that is involved in several physiological mechanisms including seed dormancy, leaf abscission, stomatal movement, and plant stress responses. Through complex interactions with several intracellular signaling systems, it can regulate the expression of hundreds of plant genes.

Other Notes

Natural isomer of the abscission-accelerating plant hormone.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML1932A8486SML0002
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

200

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

−20°C

storage temp.

room temp

storage temp.

2-8°C

application(s)

agriculture

application(s)

-

application(s)

-

application(s)

-


Still not finding the right product?

Explore all of our products under (+)-Abscisic acid


pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–6 of 6 Questions  
  1. How should Product A4906, (+)-Abscisic acid solutions be stored?

    1 answer
    1. A solution of (+)-Abscisic acid should be stored in single use aliquots at -20°C.

      Helpful?

  2. What concentration of Product A4906, (+)-Abscisic acid, is used in plant cell culture?

    1 answer
    1. (+)-Abscisic acid is used at 0.1 to 10 mg/L for plant cell culture.

      Helpful?

  3. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  4. How does the storage temperature relate to shipping conditions?

    1 answer
    1. The storage conditions that a Sigma-Aldrich catalog and label recommend for products are deliberately conservative. For many products, long-term storage at low temperatures will increase the time during which they are expected to remain in specification and therefore are labeled accordingly. Where short-term storage, shipping time frame, or exposure to conditions other than those recommended for long-term storage will not affect product quality, Sigma-Aldrich will ship at ambient temperature. The products sensitive to short-term exposure to conditions other than their recommended long-term storage are shipped on wet or dry ice. Ambient temperature shipping helps to control shipping costs for our customers. At any time, our customers can request wet- or dry-ice shipment, but the special handling is at customer expense if our product history indicates that the product is stable for regular shipment.

      Helpful?

  5. What solvents can be used with Product A4906, (+)-Abscisic acid?

    1 answer
    1. Abscisic acid, Product No. A4906, is soluble in ethanol (10 mg/mL), yielding a clear, colorless solution. For plant cell culture, it can be solubilized in a small amount of 1 N NaOH and diluted with water or medium.

      Helpful?

  6. How is Product A4906, (+)-Abscisic acid, prepared?

    1 answer
    1. This product is synthetic. This isomer is isolated from the racemic mixture.

      Helpful?

Reviews

No rating value

Active Filters