Skip to Content
Merck
All Photos(1)

Key Documents

A4848

Sigma-Aldrich

Amyloid β-Protein Fragment 17-40

≥87% (HPLC), lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥87% (HPLC)

form

lyophilized powder

mol wt

2393

composition

Peptide content, ≥75%

solubility

H2O: soluble

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@@H](NC(=O)[C@H](C)NC(=O)CNC(=O)[C@H](CCCCN)NC(=O)[C@H](CC(N)=O)NC(=O)[C@H](CO)NC(=O)CNC(=O)[C@@H](NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](C)NC(=O)[C@H](Cc1ccccc1)NC(=O)[C@H](Cc2ccccc2)NC(=O)[C@@H](NC(=O)[C@@H](N)CC(C)C)C(C)C)C(C)C)[C@@H](C)CC)C(=O)NCC(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCSC)C(=O)N[C@@H](C(C)C)C(=O)NCC(=O)NCC(=O)N[C@@H](C(C)C)C(=O)N[C@@H](C(C)C)C(O)=O

InChI

1S/C110H178N26O31S/c1-22-61(17)90(106(162)118-51-80(141)121-71(43-55(5)6)99(155)125-70(39-41-168-21)97(153)132-85(56(7)8)104(160)116-48-78(139)114-49-82(143)130-87(58(11)12)108(164)134-89(60(15)16)110(166)167)136-109(165)91(62(18)23-2)135-93(149)63(19)119-79(140)50-115-95(151)68(36-30-31-40-111)124-101(157)74(46-77(113)138)127-103(159)76(53-137)122-81(142)52-117-105(161)86(57(9)10)133-102(158)75(47-84(146)147)128-96(152)69(37-38-83(144)145)123-92(148)64(20)120-98(154)72(44-65-32-26-24-27-33-65)126-100(156)73(45-66-34-28-25-29-35-66)129-107(163)88(59(13)14)131-94(150)67(112)42-54(3)4/h24-29,32-35,54-64,67-76,85-91,137H,22-23,30-31,36-53,111-112H2,1-21H3,(H2,113,138)(H,114,139)(H,115,151)(H,116,160)(H,117,161)(H,118,162)(H,119,140)(H,120,154)(H,121,141)(H,122,142)(H,123,148)(H,124,157)(H,125,155)(H,126,156)(H,127,159)(H,128,152)(H,129,163)(H,130,143)(H,131,150)(H,132,153)(H,133,158)(H,134,164)(H,135,149)(H,136,165)(H,144,145)(H,146,147)(H,166,167)/t61-,62-,63-,64-,67-,68-,69-,70-,71-,72-,73-,74-,75-,76-,85-,86-,87-,88-,89-,90-,91-/m0/s1

InChI key

QKAFHCYMOBODRG-FZSVSODXSA-N

Gene Information

human ... APP(351)

Amino Acid Sequence

Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val

Application

Aβ fragment used to study aggregation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L K Simmons et al.
Molecular pharmacology, 45(3), 373-379 (1994-03-01)
Amyloid beta peptide (A beta), the major protein constituent of senile plaques in patients with Alzheimer's disease, is believed to facilitate the progressive neurodegeneration that occurs in the latter stages of this disease. Early attempts to characterize the structure-activity relationship

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service