Skip to Content
Merck
All Photos(3)

Key Documents

A4142

Sigma-Aldrich

Azaserine

≥98% (TLC)

Synonym(s):

O-Diazoacetyl-L-serine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7N3O4
CAS Number:
Molecular Weight:
173.13
Beilstein:
1726602
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Quality Level

Assay

≥98% (TLC)

form

powder

color

off-white to yellow-green

antibiotic activity spectrum

fungi

Mode of action

enzyme | inhibits

storage temp.

−20°C

SMILES string

N[C@@H](COC(=O)C=[N+]=[N-])C(O)=O

InChI

1S/C5H7N3O4/c6-3(5(10)11)2-12-4(9)1-8-7/h1,3H,2,6H2,(H,10,11)/t3-/m0/s1

InChI key

MZZGOOYMKKIOOX-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: amino acid derivatives

Application

Used in cell culture for the selection of HGPRT revertants.

Biochem/physiol Actions

Azaserine is an antibiotic and antifungal; it may also act as a tumor inducer. It is a structural analog of glutamine and competes with glutamine in binding to enzymes involved in purine biosynthesis. Azaserine inhibits purine biosynthesis by covalently reacting with cysteine residues in the enzyme active sites, such as in formylglycinamide ribonucleotide amidotransferase and PRPP amidotransferase. Azaserine can induce DNA damage via the formation of carboxymethylated bases and O6-methylguanine. Secretion of exo-1,3-β-glucanase and germ-tube formation of Candida albicans were inhibited by azaserine.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

S P Ram et al.
Journal of general microbiology, 130(5), 1227-1236 (1984-05-01)
Exo-(1----3)-beta-glucanase, beta-glucosidase, autolysin and trehalase were assayed in situ in Candida albicans during yeast growth, starvation and germ-tube formation. Cell viability, germ-tube formation, intracellular glucose-6-phosphate dehydrogenase and beta-glucosidase were unaffected in cells incubated in 0.1 M-HC1 for 15 min at
Kornberg, A., and Baker, T.
DNA Replication, 57-60 (1992)
Marie Pantaleon et al.
Biology of reproduction, 78(4), 595-600 (2007-11-30)
Although mouse oocytes and cleavage-stage embryos are unable to utilize glucose as a metabolic fuel, they have a specific requirement for a short exposure to glucose prior to compaction. The reason for this requirement has been unclear. In this study
Christiane Bierkamp et al.
The American journal of pathology, 165(6), 2135-2145 (2004-12-08)
The presence of gastrin and cholecystokinin-2 (CCK2) receptors in human preneoplastic and neoplastic gastrointestinal lesions suggests a role in cancer development. In addition to the growth-promoting action of gastrin, recently a role of the cholecystokinin-2/gastrin receptor (CCK2-R) modulating cellular morphology
A J Almeida et al.
Fungal genetics and biology : FG & B, 44(12), 1387-1398 (2007-05-22)
We herein report the development of a molecular toolbox for the dimorphic fungus Paracoccidioides brasiliensis, specifically a more efficient transformation and a gene expression system. We evaluated several parameters that influence Agrobacterium tumefaciens-mediated transformation (ATMT), such as co-cultivation conditions and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service