Skip to Content
Merck
All Photos(2)

Key Documents

A3422

Sigma-Aldrich

Achromopeptidase from bacteria

partially purified powder, ≥20,000 units/mg solid

Synonym(s):

ACP, Achromopeptidase

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

partially purified powder

Quality Level

specific activity

≥20,000 units/mg solid

mol wt

27 kDa

foreign activity

Collagenase, present

storage temp.

−20°C

Application

Achromopeptidase from bacteria may be used:
  • in the homogenization step for genomic DNA extraction from microbial biofilm
  • in the lysis of gram-positive strains(
  • for DNA extraction from Nocardia strains

Achromopeptidase is useful for lysis of Gram-positive bacteria that are resistant to lysozyme. Achromopeptidase, from Sigma, has been used for the isolation and molecular characterization of skin and soft-tissue bacterial infections. Product A3422 is from bacteria and is a partially purified powder containing lactose.

Biochem/physiol Actions

Achromopeptidase is a bacteriolytic enzyme and belongs to serine protease family. It is effective for the lysis of gram positive bacteria compared to lysozyme. Achromopeptidase activity is inhibited in the presence of heavy metals. Achromopeptidase based inhibition assay may be useful for screening the presence of heavy metals.
Achromopeptidase is a lysyl endopeptidase that has a molecular weight of approximately 27 kDa. It works best between pH 8.5 -9. Approximately 500-1,500 μn/ml achromopetidase can be used to lyse cells at a density of OD600 = 0.6 over 2 hours at 37 °C.

Unit Definition

One unit will lyse 0.4 μg of Micrococcus lysodeikticus per minute by turbidimetric detection at 600 nm when suspended in buffer at pH 8.0 at 37 °C.

Physical form

Partially purified powder.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microbial ecosystem analysis in root canal infections refractory to endodontic treatment
Henriques LC, et al.
Journal of Endodontics, 42(8), 1239-1245 (2016)
Metagenomic analysis of an ecological wastewater treatment plant?s microbial communities and their potential to metabolize pharmaceuticals
Balcom IN, et al.
F1000Research, 5, 844-846 (2016)
Near real-time inhibitive assay for heavy metals using achromopeptidase
Shukor MY, et al.
Indian Journal of Biotechnology, 13, 398-403 (2014)
High Intraspecific Genetic Diversity of Nocardia brasiliensis, a Pathogen Responsible for Cutaneous Nocardiosis Found in France: Phylogenetic Relationships by Using sod and hsp65 Genes
Kosova-Maali D, et al.
BioMed Research International, 2018 (2018)
Achromopeptidase for lysis of anaerobic gram-positive cocci.
Ezaki T and Suzuki S
Journal of Clinical Microbiology, 16(5), 844-846 (1982)

Protocols

To measure achromopeptidase activity, this procedure uses bacterial cells and a turbidimetric rate assay. Turbidity is measured at 600 nm using a spectrophotometer.

To measure achromopeptidase activity, this procedure uses bacterial cells and a turbidimetric rate assay. Turbidity is measured at 600 nm using a spectrophotometer.

To measure achromopeptidase activity, this procedure uses bacterial cells and a turbidimetric rate assay. Turbidity is measured at 600 nm using a spectrophotometer.

To measure achromopeptidase activity, this procedure uses bacterial cells and a turbidimetric rate assay. Turbidity is measured at 600 nm using a spectrophotometer.

Related Content

Find cell lysis and protein extraction reagents like cell lysis buffers, protein extraction kits, extraction enzymes, and other lysis enhancers for isolating and purifying proteins.

Find cell lysis and protein extraction reagents like cell lysis buffers, protein extraction kits, extraction enzymes, and other lysis enhancers for isolating and purifying proteins.

Find cell lysis and protein extraction reagents like cell lysis buffers, protein extraction kits, extraction enzymes, and other lysis enhancers for isolating and purifying proteins.

Find cell lysis and protein extraction reagents like cell lysis buffers, protein extraction kits, extraction enzymes, and other lysis enhancers for isolating and purifying proteins.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service