Skip to Content
Merck
All Photos(1)

Key Documents

A1902

Sigma-Aldrich

Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Rabbit Anti-Mouse IgG (whole molecule)−AP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

mouse

technique(s)

direct ELISA: 1:1,000
western blot: suitable

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Mouse IgG is a plasma B cell derived antibody isotype defined by its heavy chain. IgG is the most abundant antibody isotype found in mouse serum.

Specificity

Rabbit polyclonal anti-Mouse IgG (whole molecule)-Alkaline Phosphatase reacts with mouse IgA, IgG1, IgG2a, IgG2b, IgG3, and IgM myeloma proteins.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Enzyme-linked immunosorbent assay (1 paper)
Western Blotting (1 paper)
Rabbit anti-Mouse IgG (whole molecule)-alkaline phosphatase antibody has been used for ELISA and western blot assays.
Western blot analysis of lysates from the placental cell line 3ASubE-P-3 and U037 cells were performed using alkaline phosphatase conjugated rabbit anti- mouse IgG as the secondary at 1:2000 to recognize the mouse mom Il-8RB. Secondary was developed using bromochloroindoyl phosphate (Sigma) pH. 9.5 for 10-15 minutes.Western blot analysis of coimmunoprecipitates from human cell or rat cell vaginal extracts were performed using alkaline phosphatase conjugated rabbit anti- mouse IgG as the secondary at 1:15000 dilution.

Biochem/physiol Actions

IgG has an ability to cross the placental barrier. It acts as a complement activator and binds to the Fc-receptors on phagocytic cells. The level of IgG may vary with the status of disease or infection.

Physical form

Solution in 0.05 M Tris, pH 8.0, and 1 mM MgCl2 containing 1% bovine serum albumin, 50% glycerol, and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A M Robinson et al.
Biochimica et biophysica acta, 1369(2), 278-286 (1998-04-29)
Immunoliposomes have been prepared using antibodies raised to an antigenic determinant on the cell surface of the oral bacterium Streptococcus oralis (S. oralis) in an investigation of their potential to reduce dental plaque. The N-succinimidyl-S-acetylthioacetate (SATA) derivative of the antibodies
Immunohistochemical Detection of a Unique Protein within Cells of Snakes Having Inclusion Body Disease, a World-Wide Disease Seen in Members of the Families Boidae and Pythonidae
<BIG>Chang LW, et al</BIG>
PLoS ONE, 8, 2013-2013 null
Corey Rogers et al.
Nature communications, 8, 14128-14128 (2017-01-04)
Apoptosis is a genetically regulated cell suicide programme mediated by activation of the effector caspases 3, 6 and 7. If apoptotic cells are not scavenged, they progress to a lytic and inflammatory phase called secondary necrosis. The mechanism by which
IgG Placental Transfer in Healthy and Pathological Pregnancies
Palmeira P, et al.
Journal of immunology research, 2012 (2011)
Jianing Li et al.
The Journal of allergy and clinical immunology, 131(2), 442-450 (2012-11-28)
The prevalence of peanut allergies is increasing. Peanuts and many other allergen sources contain significant amounts of triglycerides, which affect absorption of antigens but have unknown effects on sensitization and anaphylaxis. We recently reported that dietary medium-chain triglycerides (MCTs), which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service