A1527
Adrenocorticotropic Hormone Fragment 7-38 human
≥97% (HPLC)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
sterility
non-sterile
Quality Level
Assay
≥97% (HPLC)
form
powder
UniProt accession no.
storage temp.
−20°C
Gene Information
human ... POMC(5443)
Amino Acid Sequence
Phe-Arg-Trp-Gly-Lys-Pro-Val-Gly-Lys-Lys-Arg-Arg-Pro-Val-Lys-Val-Tyr-Pro-Asn-Gly-Ala-Glu-Asp-Glu-Ser-Ala-Glu-Ala-Phe-Pro-Leu-Glu
Application
Adrenocorticotropic hormone fragment 7-38 human has been used as astandard in external calibration for mass spectrometry.
Biochem/physiol Actions
Adrenocorticotropic hormone (ACTH) is a polypeptide hormone thatstimulates adrenocortical activity. It is produced by the prohormoneproopiomelanocortin (POMC), which is synthesized in the corticotroph andmelanotroph cells found in the intermediate and anterior lobe of the pituitarygland and the arcuate nucleus of the hypothalamus. ACTH is also synthesized inthe skin. It plays a role in controlling steroidogenesis in the adrenal glandand prevents reactive oxygen species (ROS)-induced cell toxicity. Higher levelsof ACTH lead to skin hyperpigmentation due to the hyperstimulation ofmelanocytes, characteristic of primary adrenal insufficiency (PAI). Deficiencyof ACTH synthesis, receptors, or signaling is associated with adrenalinsufficiency.
Other Notes
ACTH fragment that is a potent inhibitor of ACTH-stimulated adenylate cyclase.
Lyophilized from 0.1% TFA in H2O
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Cell and tissue research, 272(3), 439-445 (1993-06-01)
Within two weeks, hypophysectomy induced in rats a striking decrease in the level of circulating ACTH (the concentration of which was at the limit of sensitivity of our assay system), coupled with a net reduction in the plasma corticosterone concentration
Regulatory peptides, 8(4), 305-314 (1984-07-01)
Extracts of porcine anterior pituitary contain several corticotrophic variants of ACTH 1-39. They were isolated by adsorption chromatography, ion-exchange chromatography and reverse-phase high-performance liquid chromatography. Four variants were then identified as starting and ending at positions corresponding to ACTH 1-38
Analytical chemistry, 67(21), 3971-3978 (1995-11-01)
The utility of matrix-assisted laser desorption/ionization time-of-flight (MALDI-TOF) mass spectrometry for the analysis of C-terminal peptide ladders from carboxypeptidase Y (CPY) digestions is discussed. MALDI analysis of aliquots of an optimized time-dependent CPY digestion of ACTH 7-38 fragment allowed for
The Journal of pharmacology and experimental therapeutics, 230(1), 1-6 (1984-07-01)
Synthetic bovine parathyroid hormone containing the 1-34 NH2 terminal amino acids [bPTH-(1-34)] is capable of inhibiting stimulated uterine contraction. The purpose of the present investigation is to determine whether the inhibitory action of bPTH-(1-34) is a direct action of the
The Journal of steroid biochemistry and molecular biology, 67(2), 149-152 (1999-01-07)
Corticotropin-inhibiting peptide (CIP), the 7-38 fragment of human ACTH(1-39), is known to act as an antagonist of ACTH receptors. Accordingly, CIP has been found to inhibit ACTH-stimulated glucocorticoid secretion of dispersed rat adrenocortical cells, without per se affecting the basal
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service