Skip to Content
Merck
All Photos(1)

Key Documents

A1422

Sigma-Aldrich

p-Aminohippuric acid

≥99%

Synonym(s):

4-Aminohippuric acid, N-(4-Aminobenzoyl)glycine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C6H4C(O)NHCH2COOH
CAS Number:
Molecular Weight:
194.19
Beilstein:
1213676
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Product Name

p-Aminohippuric acid, ≥99%

Assay

≥99%

form

powder

mp

199-200 °C (dec.) (lit.)

application(s)

cell analysis

storage temp.

room temp

SMILES string

Nc1ccc(cc1)C(=O)NCC(O)=O

InChI

1S/C9H10N2O3/c10-7-3-1-6(2-4-7)9(14)11-5-8(12)13/h1-4H,5,10H2,(H,11,14)(H,12,13)

InChI key

HSMNQINEKMPTIC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Renal tubules secretes para amino hippuric acid/PAH.

Application

p-Aminohippuric acid has been used as infusions to measure plasma flow.

Biochem/physiol Actions

Para-Aminohippuric acid (PAH), OAT-1 specific, is used to help differentiate and characterized organic anion transporter (OAT) isoforms.
Para-aminohippuric acid (PAH) is used as a marker in nutrition studies to measure the flow of blood in pigs, which inturn helps to determine the portal-drained viscera (PDV) flux of nutrients.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vivo measurement of nitric oxide production in porcine gut, liver and muscle during hyperdynamic endotoxaemia
Bruins MJ, et al.
British Journal of Pharmacology, 137(8), 1225-1236 (2002)
Glomerular Filtration
Quantitative Human Physiology, 633-641 (2012)
Influence of para-aminohippuric acid analysis on net portal-drained viscera flux of nutrients in pigs
Fernandez-FI, et al.
Animal : An International Journal of Animal Bioscience, 12(2), 232-238 (2018)
Intragastric bolus feeding of meals containing elementary, partially hydrolyzed or intact protein causes comparable changes in interorgan substrate flux in the pig
Deutz NEP, et al.
Clinical Nutrition (Edinburgh, Scotland), 15(3), 119-128 (1996)
Effect of prolonged hyperdynamic endotoxemia on jejunal motility in fasted and enterally fed pigs
Bruins MJ, et al.
Annals of Surgery, 237(1), 44-44 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service