Skip to Content
Merck
All Photos(1)

Documents

A0910

Sigma-Aldrich

Ala-D-γ-Glu-Lys-D-Ala-D-Ala

≥97% (HPLC)

Synonym(s):

Peptidoglycan pentapeptide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H36N6O8
CAS Number:
Molecular Weight:
488.54
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26

Assay

≥97% (HPLC)

form

powder

color

white

storage temp.

−20°C

SMILES string

CC(N)C(=O)NC(CCC(=O)NC(CCCCN)C(=O)NC(C)C(=O)NC(C)C(O)=O)C(O)=O

InChI

1S/C20H36N6O8/c1-10(22)16(28)26-14(20(33)34)7-8-15(27)25-13(6-4-5-9-21)18(30)23-11(2)17(29)24-12(3)19(31)32/h10-14H,4-9,21-22H2,1-3H3,(H,23,30)(H,24,29)(H,25,27)(H,26,28)(H,31,32)(H,33,34)

InChI key

MCIBYIIODNXVSL-UHFFFAOYSA-N

Amino Acid Sequence

Ala-Glu-Lys-Ala-Ala

Application

Ala-D-γ-Glu-Lys-D-Ala-D-Ala is the pentapeptide tail of the peptidoglycan precursor UDPMurNAc-l-Ala-γ-d-Glu-l-Lys(Gly)(5)-d-Ala-d-Ala. Ala-D-γ-Glu-Lys-D-Ala-D-Ala is used to study the role of this peptide stem in peptidoglycan biosynthesis, degradation and function.

Biochem/physiol Actions

L-Ala-D-Glu-gamma-L-Lys-D-Ala-D-Ala, peptidoglycan pentapeptide, is the antigenic peptide determinant of peptidoglycan useful in studies of mechanisms of action of glycopeptide antibiotics.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Immunochemistry of a synthetic peptidoglycan-precursor pentapeptide.
A R Zeiger et al.
Biochemistry, 12(18), 3387-3394 (1973-08-28)
G Folena-Wasserman et al.
Journal of chromatography, 392, 225-238 (1987-04-17)
An affinity support was designed to facilitate the isolation and purification of glycopeptide antibiotics by mimicking their known affinity for the bacterial cell wall. Members of this class of antibiotics inhibit peptidoglycan biosynthesis by specifically binding to pentapeptide precursors terminating
Lorena Abadía Patiño et al.
Antimicrobial agents and chemotherapy, 49(4), 1419-1425 (2005-03-29)
Enterococcus faecalis BM4405-1, a susceptible derivative of the VanE-type vancomycin-resistant E. faecalis strain BM4405, was obtained after growth in the presence of novobiocin, an inhibitor of the GyrB subunit of DNA gyrase. In contrast to findings for BM4405, UDP-MurNAc-L-Ala-gamma-D-Glu-L-Lys-D-Ala-D-Ala (pentapeptide[D-Ala])
Sridhar Peddi et al.
Biochemistry, 48(24), 5731-5737 (2009-05-06)
Penicillin-binding proteins (PBPs) are bacterial enzymes involved in the final stages of cell wall biosynthesis and are the lethal targets of beta-lactam antibiotics. Despite their importance, their roles in cell wall biosynthesis remain enigmatic. A series of eight substrates, based
B Heymer et al.
Journal of immunology (Baltimore, Md. : 1950), 117(1), 23-26 (1976-07-01)
A radioimmunoassay has been developed for the measurement of antibodies to peptidoglycan in human sera including patients with rheumatic fever and juvenile rheumatoid arthritis. The assay is based on the percentage of binding of the hapten 125I-L-Ala-gamma-d-Glu-L-Lys-D-Ala-D-Ala, the major peptide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service