Skip to Content
Merck
All Photos(1)

Key Documents

A0162

Sigma-Aldrich

Anti-Mouse Polyvalent Immunoglobulins (G,A,M)−Alkaline Phosphatase antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Goat Anti-Mouse Polyvalent Immunoglobulins (G,A,M)−AP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse

technique(s)

direct ELISA: 1:3,000 using IgG, IgA, IgM
western blot: 1:50 dilution

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Related Categories

General description

Immunoglobulins are proteins produced by B cells in response to antigen and regulate response to bacteria and viruses. IgG is known to regulate complement fixation and placental transport. IgA has a crucial role in mucosal immunity as it restricts pathogens from entering the mucosal membrane. IgM is the largest antibody having a pentameric structure which modulates polyreactivity and removes apoptotic cells.

Application

Alkaline-phosphatase-conjugated anti-mouse polyvalent immunoglobulin was used to detect antibodies from hybridomas by indirect ELISA. The antibody was incubated in microtiter plates for 1 hour at 37°C and detected using p-nitrophenyl phosphate substrate (Sigma). Alkaline-phosphatase-conjugated anti-mouse polyvalent immunoglobulin was used to detect the reactivity of fungal cytoplasmic protein with proteins from the serum of mice infected with yeast-form Y cells by western blot analysis. The antibody was used at a 1:50 dilution.

Physical form

Solution in 0.05 M Tris, pH 8.0, containing 1% bovine serum albumin, 1 mM MgCl2 and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Santiago Cerrizuela et al.
Mechanisms of development, 154, 219-239 (2018-08-08)
The neural crest (NC) is a multipotent migratory embryonic population that is formed during late gastrulation and gives rise to a wide array of derivatives, including cells from the peripheral nervous system (PNS), the craniofacial bones and cartilages, peripheral glial
Julien de Lorgeril et al.
Nature communications, 9(1), 4215-4215 (2018-10-13)
Infectious diseases are mostly explored using reductionist approaches despite repeated evidence showing them to be strongly influenced by numerous interacting host and environmental factors. Many diseases with a complex aetiology therefore remain misunderstood. By developing a holistic approach to tackle
C Tartera et al.
Applied and environmental microbiology, 56(5), 1397-1399 (1990-05-01)
Monoclonal antibodies provide a rapid and specific means of direct detection of microorganisms in water and food samples. However, monoclonal antibodies specific for some bacteria are difficult to obtain; a good example of such a bacterium is Escherichia coli. Gnotobiotic
Özlem Ertekin et al.
Molecules (Basel, Switzerland), 23(11) (2018-11-18)
Immunoanalytical methods are frequently employed in the detection of hazardous small molecular weight compounds. However, antibody development for these molecules is a challenge, because they are haptens and cannot induce a humoral immune response in experimental animals. Immunogenic forms of
Karin Säljö et al.
International journal of molecular sciences, 22(19) (2021-10-14)
Medullary thyroid carcinoma (MTC) accounts for only 1-2% of thyroid cancers; however, metastatic MTC is a mortal disease with no cure. In this study, glycosphingolipids were isolated from human MTCs and characterized by mass spectrometry and binding of carbohydrate recognizing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service