Skip to Content
Merck
All Photos(4)

Documents

93221-001

Sigma-Aldrich

RNAstable®

Tube Kit, for room temp preservation of RNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

grade

for molecular biology

form

dry

usage

25 reactions

storage temp.

room temp

General description

SampleMatrix® by Biomatrica® is a proprietary core technology to stabilize and protect biological materials at room temperature without degradation. The SampleMatrix technology was designed by combining extremophile biology that enables long-term survival of organisms in extremely dry environments, with synthetic chemistry.

RNAstable® preserves RNA samples at room temperature. Each tube or plate contains RNAstable® as a coating at the bottom of the tube or well, which protects picogram to microgram amounts of RNA. This medium is completely dissolvable and ensures total and quantitative sample recovery.

Features and Benefits

  • Preserves RNA at ambient temperatures without degradation
  • Eliminates the need to store RNA in freezers or to ship on dry ice
  • Concentrates RNA more effectively than other methods
  • Compatible with most downstream applications

Packaging

  • (25) Tubes RNAstable
  • 1.5 mL screw-cap microfuge tube made of durable polypropylene
  • (1) tube of sterile water
  • Quick Reference Protocol
  • Barcode-labeled for sample tracking

Legal Information

Biomatrica is a registered trademark of Biomatrica
RNAstable is a registered trademark of Biomatrica
SampleMatrix is a registered trademark of Biomatrica

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gilberto E Hernandez et al.
BioTechniques, 47(2), 667-668 (2009-09-10)
RNA integrity is a critical factor in obtaining meaningful gene expression data. Current methodologies rely on maintaining samples in cold environments during collection, transport, processing, and storage procedures, which are also extremely time-sensitive. Several RNA storage products are commercially available
Wen Huang et al.
PloS one, 7(1), e30244-e30244 (2012-02-01)
In this study, mRNA-Seq was used to characterize and compare the leukocyte transcriptomes from two taurine breeds (Holstein and Jersey), and one indicine breed (Cholistani). At the genomic level, we identified breed-specific base changes in protein coding regions. Among 7,793,425
Stephen K Gire et al.
Science (New York, N.Y.), 345(6202), 1369-1372 (2014-09-13)
In its largest outbreak, Ebola virus disease is spreading through Guinea, Liberia, Sierra Leone, and Nigeria. We sequenced 99 Ebola virus genomes from 78 patients in Sierra Leone to ~2000× coverage. We observed a rapid accumulation of interhost and intrahost
Daniel S Stevens et al.
Journal of virological methods, 182(1-2), 104-110 (2012-03-22)
Two approaches to stabilize viral nucleic acid in processed clinical specimens were evaluated. HIV-1 RNA extracted from clinical specimens was stabilized in a dry matrix in a commercial product (RNAstable, Biomatrica, San Diego, CA, USA) and in a reverse-transcription reaction
Adrian Puren et al.
The Journal of infectious diseases, 201 Suppl 1, S27-S36 (2010-03-17)
RNA remains the most informative and accurate biomarker for human immunodeficiency virus type 1 load diagnostics and for surveillance of drug resistance markers. Viral load testing by nucleic acid amplification currently is a complex and expensive test that is restricted

Articles

RNAstable® preserves RNA via anhydrobiosis principles, enabling ambient temp storage and shipping.

RNAstable® preserves RNA via anhydrobiosis principles, enabling ambient temp storage and shipping.

RNAstable® preserves RNA via anhydrobiosis principles, enabling ambient temp storage and shipping.

RNAstable® preserves RNA via anhydrobiosis principles, enabling ambient temp storage and shipping.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service