Skip to Content
Merck
All Photos(1)

Key Documents

91957

Sigma-Aldrich

Monosodium methylphosphonate

99.0-101.0% (T)

Synonym(s):

Methanephosphonic acid monosodium salt, Methylphosphonic acid monosodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH4O3PNa
CAS Number:
Molecular Weight:
118.00
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.25
grade:
for analytical purposes
form:
crystals
solubility:
water: 3.54 g/30 mL, colorless

grade

for analytical purposes

Quality Level

Assay

99.0-101.0% (T)

form

crystals

concentration

18.5-20.5% Na

loss

≤2.0% loss on drying

pH

4.5-5.1

solubility

water: 3.54 g/30 mL, colorless

cation traces

Al: ≤5 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

UV absorption

λ: 260 nm Amax: ≤0.2
λ: 280 nm Amax: ≤0.07

suitability

no residue for filter test

SMILES string

O=P(C)(O[Na])O

InChI

1S/CH5O3P.Na/c1-5(2,3)4;/h1H3,(H2,2,3,4);/q;+1/p-1

InChI key

CZVWTNTXBUVAFR-UHFFFAOYSA-M

Application

Buffer component for ERLIC technique for Phosphoproteome analysis and inhibitor of protein phosphatases.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yogesh Surendranath et al.
Journal of the American Chemical Society, 134(14), 6326-6336 (2012-03-08)
The mechanism of nucleation, steady-state growth, and repair is investigated for an oxygen evolving catalyst prepared by electrodeposition from Co(2+) solutions in weakly basic electrolytes (Co-OEC). Potential step chronoamperometry and atomic force microscopy reveal that nucleation of Co-OEC is progressive
Marta Kwiatkowska et al.
Pesticide biochemistry and physiology, 109, 34-43 (2014-03-04)
The toxicity of herbicides to animals and human is an issue of worldwide concern. The present study was undertaken to evaluate toxic potential of widely used pesticide - glyphosate, its metabolites: aminomethylphosphonic acid (AMPA); methylphosphonic acid and its impurities: N-(phosphonomethyl)iminodiacetic
Mostafa Zarei et al.
Journal of proteome research, 11(8), 4269-4276 (2012-07-10)
In large-scale phosphoproteomics studies, fractionation by strong cation exchange (SCX) or electrostatic repulsion-hydrophilic interaction chromatography (ERLIC) is commonly used to reduce sample complexity, fractionate phosphopeptides from their unmodified counterparts, and increase the dynamic range for phosphopeptide identification. However, these procedures
Mostafa Zarei et al.
Journal of proteome research, 10(8), 3474-3483 (2011-06-21)
Reversible phosphorylations play a critical role in most biological pathways. Hence, in signaling studies great effort has been put into identification of a maximum number of phosphosites per experiment. Mass spectrometry (MS)-based phosphoproteomics approaches have been proven to be an
L P Wackett et al.
Journal of bacteriology, 169(2), 710-717 (1987-02-01)
Carbon-phosphorus bond cleavage activity, found in bacteria that utilize alkyl- and phenylphosphonic acids, has not yet been obtained in a cell-free system. Given this constraint, a systematic examination of in vivo C-P lyase activity has been conducted to develop insight

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service