Skip to Content
Merck
All Photos(1)

Key Documents

905H-0

Sigma-Aldrich

Cytokeratin 5 (EP1601Y) + Cytokeratin 14 (LL002)

Mouse and Rabbit Monoclonal Antibody

Synonym(s):

Cytokeratin 5 (EP1601Y) + Cytokeratin 14 (LL002) Mouse and Rabbit Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse
rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

EP1601Y + LL002, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

bottle of 1.0 mL predilute (905H-07)
bottle of 7.0 mL predilute (905H-08)

manufacturer/tradename

Cell Marque

concentration

0.1-1.5 μg/mL (predilute)

isotype

IgG3
IgG

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... KRT5(3852)

General description

Cytokeratin 5 is an intermediate filament protein of 58 kD molecular wt. amongst the cytokeratin family. It is a type II (basic) cytokeratin. Antibodies to this protein identify basal cells of squamous and glandular epithelia, myoepithelia, and mesothelium. Cytokeratin 14 is a 50 kD polypeptide found in basal cells of squamous epithelia, some glandular epithelia, myoepithelium, and mesothelial cells. Anti-Cytokeratin 5 has been useful in the differential diagnosis of metastatic carcinoma in the pleura versus epithelial mesothelioma. Almost all squamous cell carcinomas, half of transitional carcinomas, and many undifferentiated large cell carcinomas immunostain with CK5. Anti-CK14 has been demonstrated to be useful in differentiating squamous cell carcinomas from other epithelial tumors. Anti-CK5, along with anti-CK14, has found application in identifying the basaloid phenotype of breast carcinoma, a tumor with poor prognosis.

Quality


IVD

IVD

IVD

RUO

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and format
Note: This requires a keycode which can be found on your packaging or product label.

Download the latest released IFU
Note: This IFU may not apply to your specific product lot.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diagnostic Immunohistochemistry: Theranostic and Genomic Applications, 212-212 (2014)
Diagnostic Immunohistochemistry: Theranostic and Genomic Applications, 728-729 (2014)
Diagnostic Immunohistochemistry: Theranostic and Genomic Applications, 212-212 (2014)
Diagnostic Immunohistochemistry: Theranostic and Genomic Applications, 728-729 (2014)
David J Dabbs et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 19(11), 1506-1511 (2006-08-31)
The basal phenotype of breast carcinoma was demonstrated from a study of gene expression profiles, which demonstrated five carcinoma phenotypes with differing immunohistologic profiles and outcomes. The basal phenotype, so-named because of an immunohistologic profile that is similar to myoepithelial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service