Skip to Content
Merck
All Photos(1)

Key Documents

07-1402-I

Sigma-Aldrich

Anti-GLUT-2

from rabbit, purified by affinity chromatography

Synonym(s):

Solute carrier family 2, facilitated glucose transporter member 2, Glucose transporter type 2, liver, GLUT-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human, mouse

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... Slc2A2(20526)

General description

Solute carrier family 2, facilitated glucose transporter member 2 (UniProt P14246; also known as Glucose transporter type 2, liver, GLUT-2) is encoded by the Slc2a2 (also known as Glut-2, Glut2) gene (Gene ID 20526) in murine species. GLUTs (Glucose transporters) are a family of integral membrane glycoproteins that facilitative glucose uptake by cells. Molecular cloning of glucose transporters have identified a family of closely related genes that encode at least 7 proteins exhibiting high degree of homology (45% to 65%). GLUT2 is a facilitative glucose transporter that mediates the bidirectional transfer of glucose across the hepatocyte membrane. It is present at high levels in the plasma membrane of pancreatic beta cells where it initiates the first step in glucose-stimulated insulin secretion. Due to its low affinity and high capacity, GLUT2 transports dietary sugars, glucose, fructose and galactose in a large range of physiological concentrations, displaying large bidirectional fluxes in and out the cells. It also participates in the transport of glucose in small intestine and kidney along with the Na+/glucose cotransporter. Hepatic GLUT2 expression is upregulated in a dose-dependent manner according to glucose concentration in primary hepatocytes and hepatocyte cell lines. Also, hepatic GLUT2 expression is reduced during starvation and returns to normal levels after refeeding. In diabetic animals, hepatic GLUT2 expression is increased, but is corrected to normal levels when hyperglycemic conditions are corrected by exogenous insulin.

Ref.: Burkhardt, BR., et al. (2005). FEBS Lett. 579, 5759 5764.

Specificity

This polyclonal antibody targets a region within the first extracellular loop of human, mouse, and rat GLUT-2.

Immunogen

Epitope: domain of:
KLH-conjugated linear peptide corresponding to a 16-a.a. sequence within the first extracellular loop of mouse GLUT-2.

Application

Detect GLUT-2 using this rabbit polyclonal Anti-GLUT-2 purified Antibody, Cat. No. 07-1402-I, validated for use in Western Blotting.
Research Category
Signaling
Western Blotting Analysis: 1 µg/mL from a representative lot detected GLUT-2 in 10 µg of human (A431, HEK293, HeLa, HepG2, HUVEC, Jurkat, PC3), mouse (C2C12, NIH/3T3, Raw 264.7), rat (L6, PC12) cell lysates and mouse brain tissue homogenate.

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected GLUT-2 in 10 µg of HEK293 cell lysate.

Target description

~50 kDa oberved. 57.49/57.11/57.09 kDa (human/mouse/rat) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity purified.
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Max Hahn et al.
Communications biology, 3(1), 541-541 (2020-10-02)
Mouse models of Streptozotocin (STZ) induced diabetes represent the most widely used preclinical diabetes research systems. We applied state of the art optical imaging schemes, spanning from single islet resolution to the whole organ, providing a first longitudinal, 3D-spatial and
Xue Liu et al.
BMC biology, 21(1), 150-150 (2023-07-05)
Biological aging is an important factor leading to the development of pathologies associated with metabolic dysregulation, including type 2 diabetes, cancer, cardiovascular and neurodegenerative diseases. Telomere length, a central feature of aging, has additionally been identified as inversely associated with
Yumiko Takahashi et al.
Nutrients, 13(7) (2021-07-03)
We examined the effect of dietary carbohydrate intake on post-exercise glycogen recovery. Male Institute of Cancer Research (ICR) mice were fed moderate-carbohydrate chow (MCHO, 50%cal from carbohydrate) or high-carbohydrate chow (HCHO, 70%cal from carbohydrate) for 10 days. They then ran
Jessica L Huang et al.
The Journal of endocrinology, 246(1), 69-78 (2020-05-06)
There is great interest in generating functionally mature beta cells from stem cells, as loss of functional beta cell mass contributes to the pathophysiology of diabetes. Identifying markers of beta cell maturity is therefore very helpful for distinguishing stem cells
Atcharaporn Ontawong et al.
Biomedical reports, 15(3), 73-73 (2021-08-19)
The major constituents of Coffea arabica (coffee), including caffeine, chlorogenic acid and caffeic acid, exhibit antihyperglycemic properties in in vitro and in vivo models. However, whether Coffea arabica bean extract (CBE) regulates glucose uptake activity and the underlying mechanisms involved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service