Skip to Content
Merck

Skip To

80612

Lipase from Rhizopus oryzae

powder, light brown, ≥30 U/mg

Synonym(s):

Lipase F-AP 15

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
NACRES:
NA.54
UNSPSC Code:
12352204
EC Number:
232-619-9
MDL number:
EC Number:
Specific activity:
≥30 U/mg
Biological source:
fungus (Rhizopus oryzae)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

fungus (Rhizopus oryzae)

form

powder

specific activity

≥30 U/mg

color

light brown

storage temp.

2-8°C

InChI

1S/C11H9N3O2.Na/c15-8-4-5-9(10(16)7-8)13-14-11-3-1-2-6-12-11;/h1-7,16H,(H,12,14);/q;+1/b13-9-;

InChI key

QWZUIMCIEOCSJF-CHHCPSLASA-N

General description

Research Area: CELL SIGNALING,Rhizopus oryzae lipase (ROL) is a protein synthesized in a precursor form that includes a 26-amino acid presequence, followed by a 97-amino acid prosequence attached to the N-terminal of a mature sequence consisting of 269 amino acids. ROL possesses four potential N-glycosylation sites and is characterized by nine α-helices and eight β-strands, resulting in a molecule stabilized by three disulfide bonds between residues 29–269, 40–43, and 235–244. Lipases are widely distributed in various living organisms, including animals, plants, and microbes.

Application

Lipase from Rhizopus oryzae has been used:
  • to replace rabbit gastric extract for infant in vitro digestion
  • in the two-phase lipase-catalyzed system for the enzymatic preparation of biodiesel
  • in the simulation of infant gastrointestinal digestion in vitro, focusing on the characteristics of emulsions during digestion to replicate the stomach environment and adjust the emulsion pH

Biochem/physiol Actions

Lipases play a crucial role in catalyzing the breakdown of ester bonds of triglycerides at the interface between aqueous and oily layers. The lipase produced by Rhizopus species is known for its suitability in various industrial applications. In industrial settings, lipases of microbial origin are commonly extracellular and are produced during the fermentation process. The utilization of lipases has been shown to be more effective than traditional chemical syntheses in industries such as pharmaceuticals, cosmetics, oleochemicals, detergents, and fragrances.
These enzymes are widely used to modify the structure of oils and fats, tailoring natural lipids to meet specific properties useful for food, nutrition, and cosmetic applications. Lipases from filamentous fungi families, such as Rhizopus oryzae are extensively utilized in the oil and fats industry due to their high 1,3-regioselectivity toward triglycerides, making them versatile in lipid modification. Lipases are used industrially for the resolution of chiral compounds and the transesterification production of biodiesel.

Analysis Note

enzyme activity:
the optimum temperature is 40°C, the optimum pH is 7.2 (highly active from pH 6.5-7.5)

Other Notes

1 U corresponds to the amount of enzyme which releases 1 μmol fatty acid from triglycerides per minute at pH 7.2 and 37 C (olive oil as substrate)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
623056231062316
biological source

fungus (Rhizopus oryzae)

biological source

-

biological source

fungus (Rhizopus niveus)

biological source

(from Candida rugosa)

specific activity

≥30 U/mg

specific activity

~10 U/mg

specific activity

≥1.5 U/mg

specific activity

≥2 U/mg

form

powder

form

powder (fine)

form

powder (fine)

form

powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

color

light brown

color

-

color

-

color

yellow-brown


Still not finding the right product?

Explore all of our products under Lipase from Rhizopus oryzae


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters