Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lipase from
Rhizopus oryzae
Synonym(s):
Lipase F-AP 15, Lipase from
Rhizopus arrhizus
, Triacylglycerol acylhydrolase, Triacylglycerol lipase
CAS Number:
9001-62-1
EC Number:
232-619-9
Products
Product Category
enzymes (2)
Brand
Sigma-Aldrich (2)
Biological Source
Color
Physical Form
Quality Segment
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
80612
Lipase from
Rhizopus oryzae
,
powder, light brown, ≥30 U/mg
Expand
View Pricing
62305
Lipase from
Rhizopus oryzae
,
powder (fine), ~10 U/mg
Expand
View Pricing