Skip to Content
Merck
All Photos(1)

Key Documents

75574

Sigma-Aldrich

Hyaluronic acid sodium salt from Streptococcus equi

mol wt 500,000-750,000

Synonym(s):

Sodium hyaluronate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

(Streptococcus equi)

Quality Level

form

powder

mol wt

500,000-750,000

color

white

solubility

water: ~5 g/L

suitability

conforms to structure for Infrared spectrum

shipped in

wet ice

storage temp.

−20°C

SMILES string

[Na+].N([C@H]1[C@@H](O[C@@H]([C@H]([C@@H]1O[C@@H]4O[C@@H]([C@H]([C@@H]([C@H]4O)O)O)C(=O)O)O)CO)O[C@@H]2[C@H](O[C@H]([C@@H]([C@H]2O)O)O[C@@H]3[C@H]([C@@H](O[C@@H]([C@H]3O)CO)O)NC(=O)C)C(=O)O)C(=O)C

InChI

1S/C28H44N2O23.Na/c1-5(33)29-9-18(11(35)7(3-31)47-25(9)46)49-28-17(41)15(39)20(22(53-28)24(44)45)51-26-10(30-6(2)34)19(12(36)8(4-32)48-26)50-27-16(40)13(37)14(38)21(52-27)23(42)43;/h7-22,25-28,31-32,35-41,46H,3-4H2,1-2H3,(H,29,33)(H,30,34)(H,42,43)(H,44,45);/q;+1/t7-,8-,9-,10-,11-,12-,13+,14+,15-,16-,17-,18-,19-,20+,21+,22+,25-,26+,27-,28-;/m1./s1

InChI key

YWIVKILSMZOHHF-QJZPQSOGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

suitable as substrate for hyaluronidase

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyaluronic acid hydrogel stiffness and oxygen tension affect cancer cell fate and endothelial sprouting.
Shen, Y.I., et al.
Biomaterials Science (2014)
L Y W Bourguignon et al.
Oncogene, 31(2), 149-160 (2011-06-21)
MicroRNAs are often associated with the pathogenesis of many cancers, including head and neck squamous cell carcinoma (HNSCC). In particular, microRNA-21 (miR-21) appears to have a critical role in tumor cell survival, chemoresistance and HNSCC progression. In this study, we
Jason A Burdick et al.
Advanced materials (Deerfield Beach, Fla.), 23(12), H41-H56 (2011-03-12)
Hyaluronic acid (HA), an immunoneutral polysaccharide that is ubiquitous in the human body, is crucial for many cellular and tissue functions and has been in clinical use for over thirty years. When chemically modified, HA can be transformed into many
Chien-Hsi Chen et al.
American journal of physiology. Heart and circulatory physiology, 306(7), H1078-H1086 (2014-02-11)
Intramyocardial injection of bone marrow mononuclear cells (MNCs) with hyaluronan (HA) hydrogel is beneficial to the ischemic heart in a rat model of myocardial infarction (MI). However, the therapeutic efficacy and safety must be addressed in large animals before moving
Do Young Rhee et al.
The Journal of dermatological treatment, 25(5), 448-452 (2013-06-25)
Hyaluronic acid (HA) fillers are frequently used for the correction of facial soft-tissue defects. To compare the efficacy and safety of a novel monophasic HA filler (mono-HA), and a well-studied biphasic HA filler (bi-HA), in the treatment of moderate to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service