Skip to Content
Merck
All Photos(1)

Documents

42686

Sigma-Aldrich

Hyaluronic acid sodium salt from Streptococcus equi

mol wt 30,000-50,000

Synonym(s):

Sodium hyaluronate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

(Streptococcus equi)

form

powder

mol wt

30,000-50,000

color

white

solubility

water: ~5 g/L

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C28H44N2O23.Na/c1-5(33)29-9-18(11(35)7(3-31)47-25(9)46)49-28-17(41)15(39)20(22(53-28)24(44)45)51-26-10(30-6(2)34)19(12(36)8(4-32)48-26)50-27-16(40)13(37)14(38)21(52-27)23(42)43;/h7-22,25-28,31-32,35-41,46H,3-4H2,1-2H3,(H,29,33)(H,30,34)(H,42,43)(H,44,45);/q;+1/t7-,8-,9-,10-,11-,12-,13+,14+,15-,16-,17-,18-,19-,20+,21+,22+,25-,26+,27-,28-;/m1./s1

InChI key

YWIVKILSMZOHHF-QJZPQSOGSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natalie Fiander
British journal of nursing (Mark Allen Publishing), 22(9), S26-S26 (2013-06-12)
Painful bladder syndrome (PBS) and interstitial cystitis (IC) are associated with bladder pain, increased urinary frequency, urgency and reduced quality of life. The cause is still unknown, although there are several possible hypotheses. PBS/IC may significantly impact all areas of
Abdol-Mohammad Kajbafzadeh et al.
Urology, 81(5), 1040-1045 (2013-04-24)
To report the results of a single-center experience with the endoscopic injection of Deflux as the initial minimally invasive approach in patients with vesicoureteral reflux (VUR) and coexisting obstruction at the ureteropelvic junction (UPJO) level who presented with VUR. Between
Treatment of hyaluronan accumulation ameliorates high-fat diet-induced insulin resistance in mice.
Per Fogelstrand et al.
Diabetes, 62(6), 1816-1817 (2013-05-25)
Daniela Elena Costea et al.
Cancer research, 73(13), 3888-3901 (2013-04-20)
Heterogeneity of carcinoma-associated fibroblasts (CAF) has long been recognized, but the functional significance remains poorly understood. Here, we report the distinction of two CAF subtypes in oral squamous cell carcinoma (OSCC) that have differential tumor-promoting capability, one with a transcriptome
Shenglian Yao et al.
Journal of nanoscience and nanotechnology, 13(7), 4752-4758 (2013-08-02)
Nanofibers exist ubiquitously in natural extracellular matrix (ECM) of all kinds of human tissues forming hydrated interwoven network. Electrospinning nanotechnology has been proven to be a powerful technique to fabricate controllable nanofibers mimicking the natural ECM structures. Hyaluronic acid (HA)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service