Skip to Content
Merck

Skip To

M0779

Methyl β-D-glucopyranoside

≥99% (HPLC and GC)

Synonym(s):

Methyl β-D-glucoside

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C7H14O6
CAS Number:
Molecular Weight:
194.18
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352201
EC Number:
211-909-9
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99% (HPLC and GC)

form

powder

optical activity

[α]/D -36 to -34 °, c = 4% (w/v) in water

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

impurities

<11.5% water

color

white

mp

111-113  °C

solubility

water: 100 mg/mL, clear, colorless

SMILES string

CO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

InChI

1S/C7H14O6/c1-12-7-6(11)5(10)4(9)3(2-8)13-7/h3-11H,2H2,1H3/t3-,4-,5+,6-,7-/m1/s1

InChI key

HOVAGTYPODGVJG-XUUWZHRGSA-N

Application

Methyl β-D-glucopyranoside has been used in a study to assess alternansucrase acceptor reactions with methyl hexopyranosides. It has also been used in a study to investigate the specificity of yeast Saccharomyces cerevisiae in removing carbohydrates by fermentation.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M9376M028510050
description

≥99% (HPLC and GC)

description

≥99% (GC)

description

-

description

BioXtra, ≥97.0% (HPLC)

assay

≥99% (HPLC and GC)

assay

≥99% (GC)

assay

≥98% (GC)

assay

≥97.0% (HPLC)

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

gas chromatography (GC): suitable

technique(s)

gas chromatography (GC): suitable

technique(s)

HPLC: suitable

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

solubility

water: 100 mg/mL, clear, colorless

solubility

water: 50 mg/mL, clear, colorless

solubility

water: 50 mg/mL, clear, colorless

solubility

H2O: 0.1 g/mL, clear to very faintly turbid, colorless (hot)

form

powder

form

solid

form

powder

form

powder

mp

111-113  °C

mp

-

mp

176-179  °C

mp

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters