Skip to Content
Merck

Skip To

62309

Lipase from Pseudomonas cepacia

powder, light beige, ≥30 U/mg

Synonym(s):

PCL, PS Lipase, Triacylglycerol acylhydrolase, Triacylglycerol lipase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
232-619-9
EC Number:
MDL number:
Specific activity:
≥30 U/mg
Biological source:
bacterial (Pseudomonas cepacia)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bacterial (Pseudomonas cepacia)

Quality Segment

form

powder

specific activity

≥30 U/mg

storage condition

(Tightly closed. Dry)

technique(s)

cell based assay: suitable

color

light beige

solubility

H2O: 2 mg/mL, hazy, faintly yellow

UniProt accession no.

storage temp.

2-8°C

InChI

1S/C11H9N3O2.Na/c15-8-4-5-9(10(16)7-8)13-14-11-3-1-2-6-12-11;/h1-7,16H,(H,12,14);/q;+1/b13-9-;

InChI key

QWZUIMCIEOCSJF-CHHCPSLASA-N

General description

Research area: cell-signaling

Lipase is a hydrolytic enzyme, found ubiquitously in nature. It belongs to the α/β-hydrolases fold family. Lipase structure contains amphipathic helical lid domain in the active site that helps in interfacial activation of protein.

Application

Lipases are used industrially for the resolution of chiral compounds and the transesterification production of biodiesel.

Lipase from Pseudomonas cepacian has been used to:
  • catalyze the degradation of polycaprolactone scaffold
  • catalyze the hydrolysis of Morita-Baylis-Hillman acetates during enzymatic kinetic resolution of racemic Morita-Baylis-Hillman adducts
  • as a standard for the generation of a calibration curve to determine the activity of lipase produced by microorganisms isolated from sludge derived from an urban wastewater treatment plant for ethanol production.

Biochem/physiol Actions

Lipases catalyze the hydrolysis of carboxylic ester bonds in triacylglycerols to yield glycerol and free fatty acids. Tri-, di-, and monoglycerides are hydrolyzed (in decreasing order of rate). Triacylglycerol lipases specifically hydrolyze the outer links of triacylglycerols and operate exclusively on the water-lipid interface. Lipolytic products and intermediates formed during lipolysis are involved in various cell-signaling processes. Lipases have broad substrate specificity and high enantioselectivity. This property of lipase makes it a good catalyst in organic synthesis. Lipases play a vital role in fat digestion and metabolism.

Other Notes

1 U corresponds to the amount of enzyme which liberates 1 μmol oleic acid per minute at pH 8.0 and 40 °C (triolein, Cat. No. 62314 as substrate)
Chemoenzymatic synthesis of (-)-carbocyclic 7-deazaoxetanocin G.
Note: When triacetin is used as substrate, the pH is 7.4. Incubation time: 60 minutes.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
623168061262310
description

powder, light beige, ≥30 U/mg

description

powder, yellow-brown, ≥2 U/mg

description

powder, light brown, ≥30 U/mg

description

powder (fine), ≥1.5 U/mg

biological source

bacterial (Pseudomonas cepacia)

biological source

(from Candida rugosa)

biological source

fungus (Rhizopus oryzae)

biological source

fungus (Rhizopus niveus)

specific activity

≥30 U/mg

specific activity

≥2 U/mg

specific activity

≥30 U/mg

specific activity

≥1.5 U/mg

technique(s)

cell based assay: suitable

technique(s)

analytical sample preparation: suitable

technique(s)

-

technique(s)

-

form

powder

form

powder

form

powder

form

powder (fine)

solubility

H2O: 2 mg/mL, hazy, faintly yellow

solubility

water: slightly soluble

solubility

-

solubility

-

UniProt accession no.

A0A0J5WKE3

UniProt accession no.

-

UniProt accession no.

-

UniProt accession no.

-


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters