Skip to Content
Merck
All Photos(1)

Key Documents

61764

Sigma-Aldrich

Lectin from Phaseolus vulgaris

lyophilized, powder

Synonym(s):

PHA, Red kidney bean Agglutinin

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.56

biological source

Phaseolus vulgaris

Assay

≥70% (biuret)

form

powder

quality

lyophilized

mol wt

Mr ~128000

technique(s)

tissue culture: suitable

storage temp.

2-8°C

General description

Lectins are distinct proteins or glycoproteins that have at least one non-catalytic domain which fixes reversibly to a definite mono- or oligosaccharide. Some of their potentially utilizable biological assets include: antitumor, immunomodulatory and anti-insect, antifungal, antibacterial, anti-HIV, and mitogenic properties.

Application

Lectin was used in isolation and culture of optimized peripheral blood mononuclear cells (PBMCs).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marsha Feske et al.
Clinical and vaccine immunology : CVI, 15(10), 1616-1622 (2008-08-30)
Children and immunocompromised adults are at an increased risk of tuberculosis (TB), but diagnosis is more challenging. Recently developed gamma interferon (IFN-gamma) release assays provide increased sensitivity and specificity for diagnosis of latent TB, but their use is not FDA
Arishya Sharma et al.
Journal of biomedicine & biotechnology, 2009, 929568-929568 (2009-04-04)
A lectin has been isolated from seeds of the Phaseolus vulgaris cv. "Anasazi beans" using a procedure that involved affinity chromatography on Affi-gel blue gel, fast protein liquid chromatography (FPLC)-ion exchange chromatography on Mono S, and FPLC-gel filtration on Superdex
Lok R Joshi et al.
Frontiers in immunology, 12, 747574-747574 (2021-11-23)
Swine influenza is a highly contagious respiratory disease of pigs caused by influenza A viruses (IAV-S). IAV-S causes significant economic losses to the swine industry and poses challenges to public health given its zoonotic potential. Thus effective IAV-S vaccines are
D Porter et al.
Frontiers in immunology, 13, 1023235-1023235 (2022-11-08)
The use of functional feeds for farmed fish is now regarded as a key factor in improving fish health and performance against infectious disease. However, the mechanisms by which these nutritional components modulate the immune response are not fully understood.
Sunita Yadav et al.
Cellular immunology, 361, 104272-104272 (2021-01-15)
Visceral leishmaniasis (VL) is a potentially fatal parasitic disease causing high morbidity and mortality in developing countries. Vaccination is considered the most effective and powerful tool for blocking transmission and control of diseases. However, no vaccine is available so far

Articles

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service