Skip to Content
Merck
All Photos(1)

Key Documents

61743

Sigma-Aldrich

N-Lauroylsarcosine sodium salt

BioUltra, for molecular biology, ≥99.0% (HPLC)

Synonym(s):

N-lauroylsarcosine sodium salt, N-Dodecanoyl-N-methylglycine sodium salt, Sarkosyl NL

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10CON(CH3)CH2COONa
CAS Number:
Molecular Weight:
293.38
Beilstein:
5322974
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

grade

for molecular biology

Quality Level

description

anionic

product line

BioUltra

Assay

≥99.0% (HPLC)

form

powder

mol wt

micellar avg mol wt 600
average mol wt 600

aggregation number

2

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected

loss

≤1% loss on drying, 110 °C

pH

7.0-9.0 (25 °C, 1 M in H2O)

CMC

14.6 mM (20-25°C)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.2
λ: 280 nm Amax: 0.06

SMILES string

[Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O

application(s)

sample preservation

InChI

1S/C15H29NO3.Na/c1-3-4-5-6-7-8-9-10-11-12-14(17)16(2)13-15(18)19;/h3-13H2,1-2H3,(H,18,19);/q;+1/p-1

InChI key

KSAVQLQVUXSOCR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

The N-lauroylsarcosine sodium salt is applicable for processes that require tight control of elemental content. This product aids in the solubilization and separation of membrane proteins, glycoproteins, and the isolation of RNA and plasmids. N-lauroylsarcosine sodium salt serves as an antifoaming supplement or as a component of a lysing buffer of bacteria or other cells. It indicates the paramagnetic anisotropy sign change in the micelle mesophase. N-lauroylsarcosine sodium salt inhibits hexokinase and the bacterial flora of human saliva/gut at 0.25%. It also exhibits fungistatic activity in aqueous dispersion (1%). N-lauroylsarcosine (LS) is an anionic surfactant with protein denaturant potency that is structurally similar to sodium lauryl sulfate (SLS). It can inactivate herpes simplex virus (HSV) infectivity in vitro. It is used as a microbicide in topical vaginal formulations to block the transmission of HSV, human immunodeficiency virus type 1 (HIV-1), and possibly other pathogens causing sexually transmitted diseases (STDs).[1]

Application

N-Lauroylsarcosine sodium salt has been used as a component:

  • of nuclease stop mix, embryo extract buffer to dissociate nonspecifically bound proteins from chromatin
  • of proteinase K solution for the heat denaturation of genomic DNA
  • of 10% (w/v) sarcosyl/0.5 M ethylenediaminetetraacetic acid (EDTA) solution to prepare EDTA-sarcosyl-proteinase (ESP) buffer
  • in the hybridization buffer for in situ hybridization histochemistry[2]
  • in the embryo extract buffer to dissociate nonspecifically bound proteins from chromatin[3]
  • in proteinase K solution to inactivate the enzymes[4]

Other Notes

Anionic surfactant.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

512.6 °F - closed cup

Flash Point(C)

267 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

UV laser cross-linking: a real-time assay to study dynamic protein/DNA interactions during chromatin remodeling
Akhilesh NK and Gordon HL
Science's STKE : Signal Transduction Knowledge Environment (2004)
Ahmed I Abo-Ahmed et al.
Acta histochemica, 122(6), 151597-151597 (2020-08-12)
The retina possesses few types of neurons so; it is considered an excellent model for understanding the neural mechanisms underlying basic neural information processing in the brain. Glutamate is the major excitatory neurotransmitter in the vertebrate central nervous system and
Iryna Benilova et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(38), 23815-23822 (2020-09-10)
Prions are infectious agents which cause rapidly lethal neurodegenerative diseases in humans and animals following long, clinically silent incubation periods. They are composed of multichain assemblies of misfolded cellular prion protein. While it has long been assumed that prions are
Shunfei Yan et al.
British journal of cancer, 124(3), 616-627 (2020-11-12)
Intrinsic and acquired drug resistance represent fundamental barriers to the cure of high-grade serous ovarian carcinoma (HGSC), the most common histological subtype accounting for the majority of ovarian cancer deaths. Defects in homologous recombination (HR) DNA repair are key determinants
Carl Alexander Sandhof et al.
Autophagy, 16(5), 878-899 (2019-07-30)
Aging is associated with a gradual decline of cellular proteostasis, giving rise to devastating protein misfolding diseases, such as Alzheimer disease (AD) or Parkinson disease (PD). These diseases often exhibit a complex pathology involving non-cell autonomous proteotoxic effects, which are

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service