Skip to Content
Merck
All Photos(1)

Key Documents

57600

Sigma-Aldrich

IL-2 human

≥98% (GE), recombinant, expressed in E. coli, liquid

Synonym(s):

Interleukin-2 human, TCGF, hIL-2, IL-2

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

Product Name

Interleukin-2 human, recombinant, expressed in E. coli, ~10000 U/mL

biological source

human

recombinant

expressed in E. coli

form

liquid

mol wt

Mr ~15500

packaging

vial of 1 mL

concentration

~10000 U/mL

color

colorless

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... IL2(3558)

Biochem/physiol Actions

Promotes the growth of IL-2 dependent lymphocytes. Amino acid sequence of IL-2.

Packaging

One vial contains 10′000 Units

Caution

Stable at −20 °C; prepare appropriate aliquots and avoid repeated freezing and thawing, since aggregation may occur resulting in apparent loss of activity.

Unit Definition

1 U corresponds to the unit activity in the colorimetric MTT-assay with CTLL-2 cells.

Physical form

Clear; colorless solution in Dulbecco-PBS; containing 1 mg BSA/mL.

Other Notes

Sales restrictions may apply.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interleukin 2.
K A Smith
Annual review of immunology, 2, 319-333 (1984-01-01)
S Gillis et al.
Journal of immunology (Baltimore, Md. : 1950), 120(6), 2027-2032 (1978-06-01)
Several soluble factors have recently been associated with the proliferation and differentiation of thymus-derived lymphocytes. One of these factors present in medium conditioned by T cell mitogen-stimulated lymphocytes has the ability to promote the long-term culture of normal and antigen-specific
Sagar Darvekar et al.
Cancers, 12(2) (2020-02-12)
Extracorporeal photopheresis (ECP), a modality that exposes isolated leukocytes to the photosensitizer 8-methoxypsoralen (8-MOP) and ultraviolet-A (UV-A) light, is used to treat conditions such as cutaneous T-cell lymphoma and graft-versus-host disease. However, the current procedure of ECP has limited selectivity
Cecilie Brekke Rygh et al.
PloS one, 9(9), e108414-e108414 (2014-10-01)
There are currently no established radiological parameters that predict response to immunotherapy. We hypothesised that multiparametric, longitudinal magnetic resonance imaging (MRI) of physiological parameters and pharmacokinetic models might detect early biological responses to immunotherapy for glioblastoma targeting NG2/CSPG4 with mAb9.2.27
R J Robb et al.
Proceedings of the National Academy of Sciences of the United States of America, 81(20), 6486-6490 (1984-10-01)
Human interleukin 2 was separated into multiple molecular forms by selective immunoaffinity chromatography and chromatofocusing. For the most part, this heterogeneity was attributed to variations in glycosylation of the threonine residue in position 3 of the polypeptide chain. The various

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service