50661
D-Tagatose 6-phosphate lithium salt
≥95% (TLC)
Synonym(s):
D-Tagatofuranose 6-phosphate lithium salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H13O9P · xLi+
CAS Number:
Molecular Weight:
260.14 (free acid basis)
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25
Recommended Products
Related Categories
Application
D-Tagatose 6-phophate is a metabolite in the biosynthetic pathway converting galactose 6-phosphate to pyruvate. D-Tagatose 6-phosphate is a substrate for tagatose-6-phosphate kinase which converts it into D-tagatose 1,6-bisphosphate with the transfer of a phosphate from ATP.
Biochem/physiol Actions
An enzyme substrate for D-tagatose-6-phosphate kinase, and key metabolite of the tagatose-6-phosphate pathway linking D-galactose-6-phosphate with D-glyceraldehyde-3-phosphate.
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Linda Miallau et al.
The Journal of biological chemistry, 282(27), 19948-19957 (2007-04-27)
High resolution structures of Staphylococcus aureus d-tagatose-6-phosphate kinase (LacC) in two crystal forms are herein reported. The structures define LacC in apoform, in binary complexes with ADP or the co-factor analogue AMP-PNP, and in a ternary complex with AMP-PNP and
V L Crow et al.
Journal of bacteriology, 153(1), 76-83 (1983-01-01)
The three enzymes of the D-tagatose 6-phosphate pathway (galactose 6-phosphate isomerase, D-tagatose 6-phosphate kinase, and tagatose 1,6-diphosphate aldolase) were absent in lactose-negative (Lac-) derivatives of Streptococcus lactis C10, H1, and 133 grown on galactose. The lactose phosphoenolpyruvate-dependent phosphotransferase system and
B Nobelmann et al.
Biochimica et biophysica acta, 1262(1), 69-72 (1995-05-17)
The sequence of the gat operon for galactitol (Gat) utilization from a wild-type isolate of Escherichia coli, strain EC3132, is presented. The operon comprises 7 open reading frames (ORFs) called gatYZABCDR. The genes are transcribed from a promoter located upstream
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service