Skip to Content
Merck
All Photos(1)

Key Documents

47989

Sigma-Aldrich

Fura 2-AM

BioReagent, suitable for fluorescence, ≥95.0% (HPLC)

Synonym(s):

Fura-2 pentakis(acetoxymethyl) ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H47N3O24
CAS Number:
Molecular Weight:
1001.85
Beilstein:
8183748
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Assay

≥95.0% (HPLC)

form

solid

solubility

DMSO: soluble

fluorescence

λex 355 nm; λem 495 nm in 0.1 M Tris pH 8.0, esterase; 10 mM Ca2+

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

CC(=O)OCOC(=O)CN(CC(=O)OCOC(C)=O)c1ccc(C)cc1OCCOc2cc3cc(oc3cc2N(CC(=O)OCOC(C)=O)CC(=O)OCOC(C)=O)-c4ncc(o4)C(=O)OCOC(C)=O

InChI

1S/C44H47N3O24/c1-25-7-8-32(46(16-39(53)65-20-60-26(2)48)17-40(54)66-21-61-27(3)49)35(11-25)58-9-10-59-36-12-31-13-37(43-45-15-38(71-43)44(57)69-24-64-30(6)52)70-34(31)14-33(36)47(18-41(55)67-22-62-28(4)50)19-42(56)68-23-63-29(5)51/h7-8,11-15H,9-10,16-24H2,1-6H3

InChI key

VPSRLGDRGCKUTK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fura 2-AM (Fura 2), also known as acetoxy-methyl-ester, is a calcium-specific ratio-metric fluorescent dye. In response to calcium ions, ratio metric dyes like Fura 2 changes their respective excitation or emission spectra. The intracellular calcium concentration is determined by the fluorescence emission ratio or excitation at distinct wavelengths. Fura 2 has an emission peak at 505nm, and the excitation peak ranges between 340-380nm and is the preferred calcium indicator.

Application

Fura 2-AM is a popular probe used to determine dynamic changes in cytosolic free calcium in intact living cells.

Features and Benefits

The major benefits of using the ester form of Fura 2 as Fura 2-AM are:

  • It is cell-permeable.
  • It can be loaded into cells easily.
Other advantages of using Fura 2-AM over single wavelength dyes is that the ratio signal is independent of :
  • The dye concentration.
  • Illumination intensity, and
  • Optical path length.

Other Notes

Membrane-permeable derivative of Fura-2. Used to load the chelator into cells. Hydrolysed enzymatically in the cell to yield Fura-2 which remains within the cell

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Almers et al.
FEBS letters, 192(1), 13-18 (1985-11-11)
The Ca concentration ([Ca2+]i) in single rat peritoneal mast cells was measured by means of the new fluorescent Ca-indicator dye fura-2. Dye-loaded cells were made to degranulate with either antigen or compound 48/80. In cells loaded with extracellularly applied, membrane-permeant
The use of microinjection and video microscopy for the study of calmodulin and calcium in living cells.
M Shelanski et al.
Methods in enzymology, 139, 824-834 (1987-01-01)
Rubin Gulaboski et al.
Cell calcium, 43(6), 615-621 (2007-11-16)
Fura-2 is one of the most commonly used fluorescent dyes to analyze the cytosolic Ca(2+) concentration ([Ca(2+)](i)) of living cells. Fura-2-dependent measurements of [Ca(2+)](i) are susceptible to changes of pH, reactive oxygen species concentration and membrane potential. Fura-2 is often
Robin R Hodges et al.
Experimental eye research, 103, 99-113 (2012-09-15)
The purpose of this study was to identify the signaling pathways that epidermal growth factor (EGF) uses to stimulate mucin secretion from cultured rat conjunctival goblet cells and to compare the pathways used by EGF with those used by the
M Poenie et al.
Nature, 315(6015), 147-149 (1985-05-09)
Although the regulation of events in the cell division cycle by calcium or other cations has been the subject of much interest and speculation, experimental studies have been hampered by the difficulty of measuring submicromolar intracellular free calcium concentrations ([Ca2+]i)

Articles

Fluorescence lifetime measurement is advantageous over intensity-based measurements. Applications include fluorescence lifetime assays, sensing and FLI.

Fluorescence lifetime measurement is advantageous over intensity-based measurements. Applications include fluorescence lifetime assays, sensing and FLI.

Fluorescence lifetime measurement is advantageous over intensity-based measurements. Applications include fluorescence lifetime assays, sensing and FLI.

Fluorescence lifetime measurement is advantageous over intensity-based measurements. Applications include fluorescence lifetime assays, sensing and FLI.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service