Skip to Content
Merck
All Photos(4)

Key Documents

318337

Sigma-Aldrich

Acridine Orange hydrochloride hydrate

≥98% (HPLC)

Synonym(s):

3,6-Bis(dimethylamino)acridine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19N3 · HCl · xH2O
CAS Number:
Molecular Weight:
301.81 (anhydrous basis)
Colour Index Number:
46005
Beilstein:
3734978
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

≥98% (HPLC)

form

powder

technique(s)

microbe id | staining: suitable

color

orange

mp

284-287 °C (lit.)

solubility

H2O: 0.1%, clear, orange to red

λmax

492 nm

ε (extinction coefficient)

≥14,000 at 227-233 nm in water at 0.005 g/L
≥35000 at 265-271 nm in water at 0.005 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cl[H].[H]O[H].CN(C)c1ccc2cc3ccc(cc3nc2c1)N(C)C

InChI

1S/C17H19N3.ClH.H2O/c1-19(2)14-7-5-12-9-13-6-8-15(20(3)4)11-17(13)18-16(12)10-14;;/h5-11H,1-4H3;1H;1H2

InChI key

JUAAUEXLEKAEQC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

A rigorously purified zinc-free material suitable for use in flow cytometry.
Acridine Orange hydrochloride hydrate dye has been used:
  • to stain intracellular acidic vesicles.
  • to measure lysosomal membrane permeability (LMP).
  • or autophagy detection.
Fluorescent stain for nucleic acids in agarose and polyacrylamide gels. Has been used to study apoptosis in various cell culture and tissue models.

Biochem/physiol Actions

Acridine Orange is a metachromatic dye which can stain DNA, RNA and acid glycosaminoglycans. At low concentrations, it intercalates into DNA and precipitates RNA. However, at high concentrations it denatures and precipitates both RNA and DNA. Acridine Orange is also used to analyze autophagy. It enters into acidic organelles in a pH-dependent manner. At neutral pH, acridine orange gives a green fluorescence and in acidic conditions, it accumulates in acidic organelles giving a bright red fluorescence.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Viriditoxin regulates apoptosis and autophagy via mitotic catastrophe and microtubule formation in human prostate cancer cells.
Kundu S, et.al.
International Journal of Oncology, 45, 2331-2340 (2014)
F. W. D. Rost
Fluorescence Microscopy, 2 (1992)
Autophagy inhibition uncovers the neurotoxic action of the antipsychotic drug olanzapine
Vucicevic L
Autophagy, 10, 2362-2378 (2014)
Autophagy-dependent senescence in response to DNA damage and chronic apoptotic stress
Singh K, et.al.
Autophagy, 8, 236-251 (2012)
Michael M Fisher et al.
International journal of radiation oncology, biology, physics, 107(4), 793-803 (2020-04-17)
We examined the capacity of the pan-fibroblast growth factor receptor (FGFR) inhibitor AZD4547 to augment radiation response across a panel of head and neck squamous cell carcinoma (HNSCC) cell lines and xenografts. FGFR1, FGFR2, and FGFR3 RNA in situ hybridization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service