Skip to Content
Merck
All Photos(1)

Key Documents

46119

Supelco

Pymetrozin

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11N5O
CAS Number:
Molecular Weight:
217.23
Beilstein:
7814151
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CC1=NNC(=O)N(C1)\N=C\c2cccnc2

InChI

1S/C10H11N5O/c1-8-7-15(10(16)14-13-8)12-6-9-3-2-4-11-5-9/h2-6H,7H2,1H3,(H,14,16)/b12-6+

InChI key

QHMTXANCGGJZRX-WUXMJOGZSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Graham Goldsworthy
Journal of insect physiology, 56(8), 991-997 (2010-04-27)
When injected into adult or nymphal Locusta that have been deprived of food for 2h, immunogens such as laminarin and bacterial LPS can induce an almost immediate dose-dependent state of anorexia for at least 1h. Such anorexia is a component
Charles I Abramson et al.
Ecotoxicology and environmental safety, 78, 287-295 (2011-12-27)
Experiments are designed to examine the effects of pymetrozine (Plenum WG-50), a recent systemic pesticide of the pyridine-azomethin family, on Pavlovian conditioning of harnessed foragers. In one set of experiments bees learned a task in which they associated a conditioned
Nikos Karatolos et al.
Pest management science, 66(12), 1304-1307 (2010-08-28)
Trialeurodes vaporariorum (Westwood), also known as the greenhouse whitefly, is a serious pest of protected vegetable and ornamental crops in most temperate regions of the world. Neonicotinoid insecticides are used widely to control this species, although resistance has been reported
Xian Zhang et al.
Annali di chimica, 97(5-6), 295-301 (2007-08-19)
For the first time, a novel, simple and reliable method for analysis of pymetrozine residues in flue-cured tobacco leaves has been developed utilizing HPLC-UV with liquid-liquid partition cleanup. Pre-treatment with ultrasonic extraction and liquid-liquid partition procedures gave preferable baseline separation
Hülya Mercan et al.
Journal of hazardous materials, 141(3), 700-706 (2006-09-05)
A simple, fast and sensitive differential pulse polarographic method (DPP) for the determination of pymetrozine insecticide in pure form, agrochemical formulation, natural water and orange juice samples is proposed. The polarographic behavior of pymetrozine exhibited a double well-defined polarographic peaks

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service