Skip to Content
Merck
All Photos(1)

Key Documents

434R-1

Sigma-Aldrich

ERG (EP111) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP111, monoclonal

description

(For In Vitro Diagnostic Use in Select Regions (See Chart))

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (434R-14)
vial of 0.5 mL concentrate (434R-15)
bottle of 1.0 mL predilute (434R-17)
vial of 1.0 mL concentrate (434R-16)
bottle of 7.0 mL predilute (434R-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:200

isotype

IgG

control

tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

Gene Information

human ... ERG(2078)

General description

ERG is a member of the erythroblastosis virus E26 transforming sequence (ETS) transcription factor gene family, which also includes Fli-1 and ETS-1.1-5 ERG is expressed in lymphocytes and endothelial cells and regulates endothelial apoptosis and angiogenesis. ERG has been found to be expressed in both benign and malignant vascular tumors. Anti-ERG was found to have high sensitivity for vascular neoplasms.1 ERG expression has been observed in prostate carcinomas and high-grade prostatic intraepithelial neoplasia (HGPIN).1-4 ERG positivity in any other epithelial tumor other than prostate carcinoma is extremely rare. In a study of all carcinomas of the breast, gastrointestinal tract, gynecologic system, kidney, lung, ovary, pancreas, salivary glands, skin, thyroid, and testis were negative for ERG.1 ERG has also been observed to be positive in a minority of Ewing sarcomas.2 This is caused by a chromosomal rearrangement which fuses the EWSR1 gene with the ERG gene (EWSR 1:ERG).5 ERG has also been observed to stain some meningiomas due to cross reactivity with Fli-1.1

Quality


IVD

IVD

IVD

RUO

Linkage

ERG Positive Control Slides, Product No. 434S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jason L Hornick
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 27 Suppl 1, S47-S63 (2014-01-05)
Immunohistochemistry plays a key role in the diagnosis of soft tissue tumors. Until recently, however, the primary purpose of immunohistochemistry in this context was simply to attempt to demonstrate a line of differentiation. Unfortunately, most traditional markers (predominantly directed against
Oksana Yaskiv et al.
American journal of clinical pathology, 138(6), 803-810 (2012-11-20)
Avian v-ets erythroblastosis virus E26 oncogene homolog (ERG) is highly sensitive and specific for endothelial neoplasms and specific for prostate carcinoma. We characterized a rabbit anti-ERG antibody as an immunohistochemical agent to detect ERG expression in various tumors using tissue
Markku Miettinen et al.
The American journal of surgical pathology, 35(3), 432-441 (2011-02-15)
ERG, an ETS family transcription factor, is known to be expressed in endothelial cells, and oncogenic ERG gene fusions occur in subsets of prostatic carcinoma, acute myeloid leukemia, and Ewing sarcoma. In this study, we immunohistochemically investigated nuclear ERG expression
J R Bänffer et al.
Antonie van Leeuwenhoek, 53(3), 183-190 (1987-01-01)
The immune response was studied in 238 human patients with Campylobacter jejuni/coli (CJC)-infections in Rotterdam by the counterimmunoelectrophoresis (CIE) test, a commercial complement fixation test (CFT) and the passive haemagglutination test (HA). Antibodies became detectable in the three tests around
Sarah Minner et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 26(1), 106-116 (2012-08-18)
Approximately 50% of prostate cancers are characterized by TMPRSS2 (transmembrane protease serine 2)-ERG (avian v-ets erythroblastosis virus E26 oncogene homolog) gene fusions resulting in an androgen-regulated overexpression of the transcription factor ERG. Some studies have suggested prognostic or predictive relevance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service