Skip to Content
Merck
All Photos(1)

Key Documents

42424

Sigma-Aldrich

Atto 550

for fluorescence, ≥90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352108
NACRES:
NA.32

grade

for fluorescence

Assay

≥90% (HPLC)

form

powder

manufacturer/tradename

ATTO-TEC GmbH

λ

in ethanol (with 0.1% trifluoroacetic acid)

UV absorption

λ: 553-559 nm Amax

suitability

suitable for fluorescence

storage temp.

−20°C

General description

Atto 550 is a new label with high molecular absorption (120,000) and quantum yield (0.80) as well as sufficient Stokes shift between excitation and emission maximum. Atto 550 can be used with similar excitation source and fluorescence filters as Cy3 and is characterized by a high photostability.

find more information here

Application

Atto fluorescent labels are designed for high sensitivity applications, including single molecule detection. Atto labels have rigid structures that do not show any cis-trans-isomerization. Thus these labels display exceptional intensity with minimal spectral shift on conjugation. Atto 550, which is similar to Cy3, may be useful in applications such as fluorescence resonance energy transfer (FRET) and as a tag for molecules such as secondary antibodies.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.
Cy3 is a trademark of Cytiva

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Smart-aggregation imaging for single molecule localization with SPAD cameras.
Gyongy, I.; et al.
arXiv (2016)
Antibody transfection into neurons as a tool to study disease pathogenesis.
Douglas JN, Gardner LA, et al.
Journal of Visualized Experiments, 26, 4154-4154 (2012)
Hanna Mannell et al.
Pharmaceutical research, 29(5), 1282-1294 (2012-01-26)
Site specific vascular gene delivery is a promising tool for treatment of cardiovascular diseases. By combining ultrasound mediated microbubble destruction with site specific magnetic targeting of lentiviruses, we aimed to develop a technique suitable for systemic application. The magnetic nanoparticle
Binding of Biotin to Streptavidin: A combined fluorescence correlation spectroscopy and time-resolved fluorescence study.
Stromqvist, J., et al.
The European Physical Journal - Special Topics, 199, 181-194 (2011)
Use of universal reporter primers in multiplex PCR of autosomal loci.
Favaro, E. C., et al.
Forensic Science International: Genetics Supplement Series, 3, e71- e72 (2011)

Articles

Fluorescence lifetime measurement is advantageous over intensity-based measurements. Applications include fluorescence lifetime assays, sensing and FLI.

Fluorescence lifetime measurement is advantageous over intensity-based measurements. Applications include fluorescence lifetime assays, sensing and FLI.

Fluorescence lifetime measurement is advantageous over intensity-based measurements. Applications include fluorescence lifetime assays, sensing and FLI.

Fluorescence lifetime measurement is advantageous over intensity-based measurements. Applications include fluorescence lifetime assays, sensing and FLI.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service